Skip to Content
Merck
All Photos(1)

Key Documents

30524

Millipore

Staph ChromoSelect Agar Base, Modified

NutriSelect® Plus, suitable for microbiology

Synonym(s):

HiCrome Staphylococcus aureus Agar Base, modified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

Pricing and availability is not currently available.

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
chromogenic mixture, 5.3 g/L
lithium chloride, 5.0 g/L
peptone special, 23.0 g/L
sodium chloride, 40.0 g/L
sodium pyruvate, 4.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.2±0.2 (25 °C)

application(s)

bioburden testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Staphylococcus spp.

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF89380200GF34069091GF62112181
manufacturer/tradename

Goodfellow 906-693-14

manufacturer/tradename

Goodfellow 893-802-00

manufacturer/tradename

Goodfellow 340-690-91

manufacturer/tradename

Goodfellow 621-121-81

size

50 × 50 mm

size

-

size

150 × 150 mm

size

-

thickness

0.1 mm

thickness

0.1 mm

thickness

0.1 mm

thickness

0.1 mm

Application

Staph ChromoSelect Agar Base for the isolation and enumeration of Staphylococcus aureus.

Preparation Note

Suspend 46.15 grams in 500 ml distilled water. Heat to boiling to dissolve the medium completely. DO NOT AUTOCLAVE. Cool to 45-50°C and aseptically add rehydrated contents of 1 vial of Polymyxin B Selective supplement (Cat. No. P9602). Mix well and pour into sterile Petri plates.

Warning : Lithium Chloride is harmful. Avoid all bodily contact and inhalation of vapours. On contact with skin wash with plenty of water immediately.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
HiCrome is a trademark of HiMedia Laboratories Pvt. Ltd.
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service