Skip to Content
Merck
All Photos(1)

Key Documents

23182

Supelco

(+)-1-(9-Fluorenyl)ethyl chloroformate solution

≥18 mM in acetone, for chiral derivatization, LiChropur

Synonym(s):

(+)-FLEC solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13ClO2
CAS Number:
Molecular Weight:
272.73
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for chiral derivatization

Quality Level

product line

ChiraSelect

optical purity

enantiomeric ratio: ≥99.0:1.0 (HPLC)

quality

LiChropur

concentration

≥18 mM in acetone

technique(s)

HPLC: suitable

refractive index

n20/D 1.359

storage temp.

2-8°C

SMILES string

ClC(=O)OC(C1c2c(cccc2)c3c1cccc3)C

InChI

1S/C16H13ClO2/c1-10(19-16(17)18)15-13-8-4-2-6-11(13)12-7-3-5-9-14(12)15/h2-10,15H,1H3

InChI key

SFRVOKMRHPQYGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(+)-1-(9-Fluorenyl)ethyl chloroformate [(+)-FLEC] is a chiral derivatizing reagent. It helps in separating enantiomers of α-amino acids, as well as helps in determining its optical purity using HPLC method.

Application

(+)-FLEC may be used in enantioselective analysis of glufosinate using reversed-phase high-performance liquid chromatography system coupled with a fluorescence detection system.

Physical form

solution, 5 mg in 1 mL acetone

Other Notes

Chiral derivatizing agent for primary and secondary amino acids and amines. The derivatives are separated by reversed-phase LC with fluorescence detection for determining the enantiomeric purity

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeineb Aturki et al.
Electrophoresis, 25(4-5), 607-614 (2004-02-26)
The indirect resolution of five beta-adrenoreceptor blocking agents (propranolol, oxprenolol, pindolol, metoprolol, and atenolol) using precolumn derivatization with (+)-1-(9-fluorenyl)ethyl chloroformate (FLEC), and capillary electrochromatography (CEC) is reported. Three octadecylsilanized (ODS) silica gel-based stationary phases, differing in particle diameter and carbon
N Todoroki et al.
Journal of chromatography. B, Biomedical sciences and applications, 728(1), 41-47 (1999-06-24)
The natural occurrence of N-methyl-D-aspartate (NMDA) is limited to the foot muscle of Scapharca broughtonii; it is a well known compound for its neuroexitatory activity. This paper describes a high-performance liquid chromatographic (HPLC) method for the determination of NMDA in
Sascha Freimüller et al.
Journal of pharmaceutical and biomedical analysis, 30(2), 209-218 (2002-08-23)
An indirect enantioseparation method for robust and precise determination of D-Carnitine (D-C) in L-Carnitine (L-C) in the range of 0.1-1.0% is presented. The method is based on derivatization of Carnitine with (+)-[1-(9-fluorenyl)-ethyl]-chloroformate ((+)-FLEC). The two diastereomers are subsequently separated of
Michelle A Camerino et al.
Organic & biomolecular chemistry, 11(16), 2571-2573 (2013-03-14)
An efficient synthesis of the enantiomers of fluorenylethylchloroformate (FLEC) has been achieved that allows the routine application of the reagent for the resolution of chiral amines including unusual amino acids. The utility of the fluorenylethoxycarbonyl (Feoc) group as a chiral
[A method for separation of DL-amino acids].
T H Akiyama et al.
Tanpakushitsu kakusan koso. Protein, nucleic acid, enzyme, 40(1), 76-83 (1995-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service