Skip to Content
Merck
All Photos(6)

Key Documents

228729

Sigma-Aldrich

Sodium L-tartrate dibasic dihydrate

≥99%, ACS reagent

Synonym(s):

Sodium tartrate dibasic dihydrate, L-(+)-Tartaric acid disodium salt, Disodium tartrate dihydrate, Sodium tartrate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4Na2O6 · 2H2O
CAS Number:
Molecular Weight:
230.08
Beilstein:
6121732
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Assay

≥99%

optical activity

[α]20/D +26°, c = 1 in H2O

impurities

≤0.005% insolubles

loss

15.61-15.71% loss on drying

pH

7.0-9.0

anion traces

chloride (Cl-): ≤5 ppm
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.01%
Fe: ≤0.001%
NH4+: ≤0.003%
heavy metals: ≤5 ppm

SMILES string

[Na+].[Na+].[H]O[H].[H]O[H].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

InChI

1S/C4H6O6.2Na.2H2O/c5-1(3(7)8)2(6)4(9)10;;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;2*1H2/q;2*+1;;/p-2/t1-,2-;;;;/m1..../s1

InChI key

FGJLAJMGHXGFDE-DGFHWNFOSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium L-tartrate is an organic acid mainly found in grapes.

Application

Sodium L-tartrate dibasic dihydrate may be used in the preparation of solid-state M–L compounds (M = bivalent metal ions ie., Mn, Fe, Co, Ni, Cu, Zn; L = tartrate). It may also be used in the preparation of copper (II) and cobalt (II) tartrate complexes by reacting with the corresponding metal salts. These complexes can undergo thermal decomposition to form Cu and Co metal oxide nanoparticles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization and thermal behaviour on solid tartrates of some bivalent metal ions
Rodrigues EC, et al.
Thermochimica Acta, 496(1), 156-160 (2009)
Synthesis of Cu and Co metal oxide nanoparticles from thermal decomposition of tartrate complexes
Palacios-Hernandez T, et al.
Inorgorganica Chimica Acta, 392, 277-282 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service