Skip to Content
Merck
All Photos(1)

Key Documents

22520

Millipore

China Blue Lactose Agar

suitable for microbiology, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

Pricing and availability is not currently available.

sterility

non-sterile

Quality Level

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 12 g/L
casein peptone, 5 g/L
china blue, 0.375 g/L
lactose, 10 g/L
meat extract, 3 g/L
sodium chloride, 5 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbe id | utilization test: suitable
microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

bioburden testing
environmental
food and beverages
water monitoring

microbiology

General description

China Blue Lactose Agar is a selective medium for differentiating lactose fermenting and lactose non-fermenting microorganisms.

Preparation Note

Dissolve 35.5 g in 1 litre distilled water. Sterilize by autoclaving at 121°C for 15 minutes.

Other Notes

Bacterial count in milk[1]

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    E. Brandt et al.
    Milchwiss. Ber., 13, 1-1 (1963)

    Articles

    Microbiological assessment of bacteria found in spices, a potent source for food spoilage and pathogens.

    Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.

    Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.

    Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.

    See All

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service