Skip to Content
Merck
All Photos(1)

Key Documents

19013

Supelco

Nickel Standard for ICP

TraceCERT®, 10 g/L Ni in nitric acid (nominal concentration)

Synonym(s):

Nickel Calibration Solution for ICP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

10 g/L Ni in nitric acid (nominal concentration)

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application


  • Protein Purification Enhancements: Nickel ICP standards are crucial for the efficient purification of polyhistidine-tagged recombinant proteins using functionalized corundum particles, optimizing biochemistry workflows in pharmaceutical research and biotechnology (Völzke et al., 2023).

  • Photocatalytic Research Applications: Utilizing nickel ICP standards, researchers have developed hydrogen-bonded organic framework ultrathin nanosheets that significantly improve the efficiency of visible-light photocatalytic CO2 reduction, a key advancement in sustainable technology applications (Yu et al., 2022).

  • Biomedical Innovations: Nickel standards are used in the clinical evaluation of zeolite-clinoptilolite supplementation to monitor its effects on blood parameters, demonstrating the material′s bioactivity and its implications for health and disease management (Kraljević Pavelić et al., 2022).

  • Toxicity Assessment in Pharmaceuticals: The comparative toxicity of interferon beta-1a impurities, including nickel ions, was evaluated to ensure the safety and efficacy of pharmaceutical products, highlighting the critical role of nickel ICP standards in quality control and regulatory compliance (Berillo, 2022).

  • Food Safety Analysis: Nickel ICP standards facilitate the development of green methodologies like electrocolorimetric extraction for the determination of Ni(II) ions in food samples, ensuring consumer safety and regulatory compliance in the food industry (Sahragard et al., 2022).

Analysis Note

10 g/L Ni in 5% nitric acid, prepared with high purity Ni metal, HNO3 and water

Other Notes

For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service