Skip to Content
Merck
All Photos(2)

Key Documents

16922

Millipore

Vegetable Peptone for Biotechnology and Microbiology

Tryptone (vegetable)
1 of 1 reviewers received a sample product or took part in a promotion

Tryptone, Encymatically Digested, for Biotechnology and Microbiology, ≥11% Total N, ≥2.0% Amino

Synonym(s):

Peptone from vegetable, Tryptone from vegetables, Vegetable hydrolysate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85
Pricing and availability is not currently available.

Product Name

Tryptone (vegetable), suitable for microbiology, meat-free alternative

biological source

plant

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

amino-nitrogen, ≥2.5%
total nitrogen (N), ≥11%

packaging

pkg of 500 g

impurities

≤15% ash

loss

≤7.0% loss on drying

pH

5.5-7.5 (25 °C, 2% in H2O)

solubility

H2O: 2%, clear, yellow

application(s)

food and beverages
vitamins, nutraceuticals, and natural products

microbiology

Looking for similar products? Visit Product Comparison Guide

General description

Tryptone is a general-purpose growth peptone obtained from vegetable source and meat-free alternative to enzymatic digest from casein. It is used as a nitrogen source in culture media formulated for isolating and cultivating fastidious and non-fastidious bacteria, fungi, and some protozoa. It is a refined hydrolysate with high solubility and is used as a microbiological nutrient in laboratory media and fermentations. It is a high-quality source of amino acids and peptides produced by the pancreatic digestion of casein.

Application

Tryptone is recommended for laboratory media and fermentation and has been used in antibiotics, toxins, and enzyme productions. It is also used as a nutrient in vaccine production where a high free amino acid content is required. Tryptone may also be used in STGG (skim milk, tryptone, glucose, glycerol) medium at -80°C, which is used to preserve Streptococcus pneumoniae, Haemophilus influenzae, and Moraxella catarrhalis isolates for at least 3 years.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service