Skip to Content
Merck
All Photos(2)

Key Documents

16715

Supelco

Thiamphenicol

analytical standard

Synonym(s):

D-threo-2,2-Dichloro-N-(β-hydroxy-α-[hydroxymethyl]-4-[methylsulfonyl]phenethyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15Cl2NO5S
CAS Number:
Molecular Weight:
356.22
Beilstein:
2819542
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

ethanol: 50 mg/mL, clear, colorless to faintly yellow

application(s)

clinical

format

neat

storage temp.

2-8°C

SMILES string

CS(=O)(=O)c1ccc(cc1)[C@@H](O)[C@@H](CO)NC(=O)C(Cl)Cl

InChI

1S/C12H15Cl2NO5S/c1-21(19,20)8-4-2-7(3-5-8)10(17)9(6-16)15-12(18)11(13)14/h2-5,9-11,16-17H,6H2,1H3,(H,15,18)/t9-,10-/m1/s1

InChI key

OTVAEFIXJLOWRX-NXEZZACHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide
Thiamphenicol is an analog of a broad spectrum antibiotic, chloramphenicol. It shows therapeutic effects in respiratory infections, venereal diseases and bacterial prostatitis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Thiamphenicol may be used as a reference standard in the determination of thiamphenicol in honey using dispersive liquid-liquid microextraction (DLLME) coupled with high-performance liquid chromatography-variable wavelength detector (HPLC-VWD).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dispersive liquid--liquid microextraction followed by high-performance liquid chromatography as an efficient and sensitive technique for simultaneous determination of chloramphenicol and thiamphenicol in honey
Chen H, et al.
Analytica Chimica Acta, 632(1), 80-85 (2009)
[Toxicity of chloramphenicol and thiamphenicol (CAP and TAP)].
G Keiser
Haematology and blood transfusion, 21, 179-195 (1978-01-01)
J Raymond et al.
Medecine tropicale : revue du Corps de sante colonial, 64(1), 33-38 (2004-07-01)
Streptococcus pneumoniae and Haemophilus influenzae are the two main pathogens responsible for bacterial respiratory tract infections. Their antimicrobial susceptibility to antibiotics like beta-lactams, macrolides or fluoroquinolones has been largely studied, while it remains less known to other antibiotics like thiamphenicol
D Malvy et al.
Intensive care medicine, 18(8), 485-487 (1992-01-01)
A 27-year-old woman, without compromised immunodefenses, experienced a Listeria meningoencephalitis, with brainstem symptoms. The identified agent exhibited poor susceptibility to usual effective antibiotics, except for penicillins. Knowledge of past history of an allergic reaction to beta-lactam antibiotics lead to appropriate
Chloramphenicol and thiamphenicol.
J Fuglesang et al.
Antibiotics and chemotherapy, 31, 1-21 (1982-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service