Skip to Content
Merck
All Photos(1)

Key Documents

156272

Sigma-Aldrich

3,5-Dinitrobenzoyl chloride

≥96.5%

Synonym(s):

DNBC, NSC 2697

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3COCl
CAS Number:
Molecular Weight:
230.56
Beilstein:
990249
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

7.6 (vs air)

Quality Level

Assay

≥96.5%

form

solid

bp

196 °C/11 mmHg (lit.)

mp

68-69 °C (lit.)

functional group

acyl chloride
nitro

SMILES string

[O-][N+](=O)c1cc(cc(c1)[N+]([O-])=O)C(Cl)=O

InChI

1S/C7H3ClN2O5/c8-7(11)4-1-5(9(12)13)3-6(2-4)10(14)15/h1-3H

InChI key

NNOHXABAQAGKRZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Crystals of 3,5-dinitrobenzoyl chloride (DNBC) belong to the orthorhombic crystal system and space group Pna21. It is widely used reagent for the detection of alcohols.

Application

3,5-Dinitrobenzoyl chloride may be used for the quantitative estimation of hydroxyl groups in pyridine. It may be used in the synthesis of following:
  • 3,5-diamino-N-cyclopropylbenzamide
  • 3,5-dinitro-N-cyclopropylbenzamide
  • 4-(3,5-dinitrophenyl)-6-(dipropan-2-ylamino)-2H-1,3,5-thiadiazine-2-thione4


3,5-Dinitrobenzoyl chloride may also be used as derivatizing agent for the following:

  • biogenic amines present in fermented foods, which can be determined using liquid chromatography method(LC)
  • polyamines, present in biological fluids, which can be determined using high performance liquid chromatography(HPLC)

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rapid Determination of Organic Hydroxyl Groups with 3, 5,-Dinitrobenzoyl Chloride.
Robinson Jr WT, et al.
Analytical Chemistry, 33(8), 1030-1034 (1961)
Synthesis and characterization of aromatic poly (amides) based on 3, 5-diamino-N-cyclopropylbenzamide.
Tundidor-Camba A, et al.
Royal Society of Chemistry Advances, 5(29), 23057-23066 (2015)
S Wongyai et al.
Biomedical chromatography : BMC, 2(6), 254-257 (1988-11-01)
Analytical methods are described for the quantitative determination of putrescine, cadaverine, spermidine, spermine and the acetylated derivatives of spermidine and spermine in biological fluids using pre-column derivatization with either benzoyl chloride or 3,5-dinitrobenzoyl chloride, which were added to each sample
3, 5-Dinitrobenzoyl chloride.
Wang HY, et al.
Acta Crystallographica Section E, Structure Reports Online, 65(10), o2460-o2460 (2009)
A Markowska et al.
Roczniki Akademii Medycznej w Bialymstoku (1995), 42(1), 129-140 (1997-01-01)
A carbocyclic analogue of distamycin was obtained, in which the N-methylpyrrole rings were substituted by disubstituted benzene rings. Additionally, N-chloro- or N-bromoacetyl groups, displaying alkylating properties, were introduced. The synthesis, starting from 3,5-dinitrobenzoyl chloride, consisted of five stages.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service