Skip to Content
Merck
All Photos(1)

Key Documents

15269

Supelco

N,O-Bis(trimethylsilyl)acetamide

for GC derivatization, LiChropur, ≥98.5% (GC)

Synonym(s):

BSA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C[=NSi(CH3)3]OSi(CH3)3
CAS Number:
Molecular Weight:
203.43
Beilstein:
1306669
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

Assay

≥98.5% (GC)

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.417 (lit.)
n20/D 1.417

bp

71-73 °C/35 mmHg (lit.)

density

0.832 g/mL at 20 °C (lit.)

SMILES string

C\C(O[Si](C)(C)C)=N/[Si](C)(C)C

InChI

1S/C8H21NOSi2/c1-8(9-11(2,3)4)10-12(5,6)7/h1-7H3/b9-8+

InChI key

SIOVKLKJSOKLIF-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,O-Bis(trimethylsilyl)acetamide is an excellent silylation reagent. For some sterically hindered primary or secondary nitro compounds, N,O-bis(trimethylsilyl)acetamide and BSTFA achieve better results than normal silylation conditions.

Application

Learn more in the Product Information
Suitable for the derivatization of 2-amino-2-dexoxyhexoses, ecdysones, hexosamines in glycosaminoglycans, 18-hydroxycorticosteroids, 15-Oxo-prostaglandin F2α, prostaglandins A,B and E and F, F, prostaglandins and metabolites, prostaglandins Emetabolites, sugar phosphates, steroids and testosterone.

Other Notes

Powerful silylating agent
Reagent for o-trimethylsilyl-aldono-1,4-lactones, trimethylsilyl and trimethylsilyl ether.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Houben-Weyl Methods of Molecular Transformations. Heteroatom Analogues of Aldehydes and Ketones.
Aggarwal VK
Science of Synthesis Knowledge Updates, 27, 593-594 (2014)
Analysis of the prostaglandins E by glass capillary gas chromatography with electron capture detection.
M Korteweg et al.
Prostaglandins, 13(6), 1221-1223 (1977-06-01)
K. Blau et al.
Handbook of Derivatives for Chromatography (1993)
J. Heberle et al.
Silylating Agents, 2nd ed. (1995)
Anna Hjelm et al.
Applied and environmental microbiology, 81(2), 726-735 (2014-11-16)
Bacterial ghosts are empty cell envelopes of Gram-negative bacteria that can be used as vehicles for antigen delivery. Ghosts are generated by releasing the bacterial cytoplasmic contents through a channel in the cell envelope that is created by the controlled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service