Skip to Content
Merck
All Photos(1)

Key Documents

14170

Sigma-Aldrich

Sodium succinate dibasic hexahydrate

puriss. p.a., ≥98.0% (NT)

Synonym(s):

Butanedioic acid disodium salt, Succinic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCH2CH2COONa · 6H2O
CAS Number:
Molecular Weight:
270.14
Beilstein:
3921044
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98.0% (NT)
form:
solid
solubility:
H2O: soluble 270 g/L at 20 °C (completely)
Pricing and availability is not currently available.

grade

puriss. p.a.

Quality Level

Assay

≥98.0% (NT)

form

solid

solubility

H2O: soluble 270 g/L at 20 °C (completely)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

O.O.O.O.O.O.[Na+].[Na+].[O-]C(=O)CCC([O-])=O

InChI

1S/C4H6O4.2Na.6H2O/c5-3(6)1-2-4(7)8;;;;;;;;/h1-2H2,(H,5,6)(H,7,8);;;6*1H2/q;2*+1;;;;;;/p-2

InChI key

ZBTUYCUNQBRXOR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium succinate dibasic hexahydrate is hexahydrate form of sodium succinate. It is generally used as a carbon source and electron donor in electrochemistry.

Application

Sodium succinate dibasic hexahydrate is used as a precipitation agent in the synthesis of ZnO-HNBs (zinc oxide- hollow nano-baskets), that is used for the degradation of rhodamine 6G.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subhash Chandra Verma et al.
Journal of bacteriology, 194(18), 4959-4971 (2012-07-17)
The chb operon of Escherichia coli is involved in the utilization of the β-glucosides chitobiose and cellobiose. The function of chbG (ydjC), the sixth open reading frame of the operon that codes for an evolutionarily conserved protein is unknown. We
Michael L Weltzer et al.
Applied and environmental microbiology, 79(4), 1353-1358 (2012-12-25)
Environmental gradients are expected to promote the diversification and coexistence of ecological specialists adapted to local conditions. Consistent with this view, genera of phototrophic microorganisms in alkaline geothermal systems generally appear to consist of anciently divergent populations which have specialized
Angélica María Sabogal-Guáqueta et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 118, 109295-109295 (2019-09-24)
Mitochondrial dysfunction and inflammation contribute to the initiation and development of several brain pathological conditions, including Alzheimer's disease and cerebral ischemia. Linalool is an aromatic plant-derived monoterpene alcohol with reported anti-inflammatory, and anti-oxidant properties. We investigated the role of linalool
Jun Kimura et al.
Cancer science, 104(8), 1083-1090 (2013-05-18)
Pancreatic cancer has a poor prognosis after complete macroscopic resection combined with chemotherapy. Even after neoadjuvant chemotherapy, R0 resection is often not possible. Moreover, current imaging techniques cannot reliably distinguish viable cancer cells from scar tissue at the resectional margin.
Ilya B Zavodnik et al.
Life sciences, 92(23), 1110-1117 (2013-05-07)
The present study was designed for further evaluation of the biochemical mechanism of hepatic mitochondrial dysfunction under oxidative damages induced by organic hydroperoxide, tert-butyl hydroperoxide (tBHP), for estimation of the molecular targets impaired during oxidative stress, and for investigation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service