Skip to Content
Merck
All Photos(1)

Key Documents

12392

Sigma-Aldrich

Benzyl benzoate

tested according to Ph. Eur.

Synonym(s):

Benzylis benzoas, Benzoic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH2C6H5
CAS Number:
Molecular Weight:
212.24
Beilstein:
2049280
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32110502
PubChem Substance ID:
NACRES:
NA.26

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

vapor pressure

1 mmHg ( 125 °C)

form

liquid

autoignition temp.

896 °F

color

clear

refractive index

n20/D 1.568 (lit.)

bp

323-324 °C (lit.)

mp

17-20 °C (lit.)

solubility

alcohol: miscible
chloroform: miscible
diethyl ether: miscible
oil: miscible
water: insoluble

density

1.118 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

O=C(OCc1ccccc1)c2ccccc2

InChI

1S/C14H12O2/c15-14(13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10H,11H2

InChI key

SESFRYSPDFLNCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Benzyl benzoate has been used as a test compound for evaluating its acaricidal activities by comparative counts, at regular intervals, with Dermutophagoides furinue populations. It has also been used for cleaning the whole-mount lacZ stained slides for microscopic visualization under a stereomicroscope.

Biochem/physiol Actions

Benzyl benzoate is the condensation product of benzoic acid and benzyl alcohol. It is utilized for treating human scabies as well as kills house dust mite.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert Voswinckel et al.
Circulation research, 93(4), 372-379 (2003-07-26)
The biological principles that underlie the induction and process of alveolization in the lung as well as the maintenance of the complex lung tissue structure are one of the major obstacles in pulmonary medicine today. Bone marrow-derived cells have been
J H Chang et al.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology, 77(3), 187-190 (1996-09-01)
Several acaricides have become available for reducing house dust mite allergen levels. The purpose of this study was to assess whether the use of benzyl benzoate (Acarosan) provides additional benefit to the usual mite control measures including encasement of mattress
Catherine W McCollum et al.
Aquatic toxicology (Amsterdam, Netherlands), 152, 152-163 (2014-04-29)
Exposure to arsenic in its inorganic form, arsenite, causes adverse effects to many different organs and tissues. Here, we have investigated arsenite-induced adverse effects on vascular tissues in the model organism zebrafish, Danio rerio. Zebrafish embryos were exposed to arsenite
A F Kalpaklioğlu et al.
Allergy, 51(3), 164-170 (1996-03-01)
Several observations have indicated that house-dust mites (HDM) play an important role in allergic diseases. Thus, the primary form of treatment should aim at reducing exposure to HDM for these patients. Allergen-avoidance measures in homes have been demonstrated to be
Karina Caballero-Gallardo et al.
Journal of agricultural and food chemistry, 59(5), 1690-1696 (2011-02-05)
A tool for integrated pest management is the use of essential oils (EOs) and plant extracts. In this study, EOs from Tagetes lucida , Lepechinia betonicifolia , Lippia alba , Cananga odorata , and Rosmarinus officinalis , species grown in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service