Skip to Content
Merck
All Photos(1)

Key Documents

114472

Sigma-Aldrich

Cresol red

indicator grade, Dye content 95 %

Synonym(s):

o-Cresolsulfonphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18O5S
CAS Number:
Molecular Weight:
382.43
Beilstein:
343399
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

indicator grade

Quality Level

form

powder or crystals

composition

Dye content, 95%

technique(s)

titration: suitable

color

red-brown

pH

7.2-8.8, yellow to red

useful pH range

1.8-2.0, orange to yellow(acid)
7.0-8.8, yellow to violet(alkaline)

mp

290 °C

density

0.998 g/cm3

λmax

367 nm (2nd)
570 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1cc(ccc1O)C2(OS(=O)(=O)c3ccccc23)c4ccc(O)c(C)c4

InChI

1S/C21H18O5S/c1-13-11-15(7-9-18(13)22)21(16-8-10-19(23)14(2)12-16)17-5-3-4-6-20(17)27(24,25)26-21/h3-12,22-23H,1-2H3

InChI key

OBRMNDMBJQTZHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cresol Red, also known as o-cresol red, belongs to the phthalein and sulphonphthalein group of dyes. Cresol red changes color from orange to yellow at pH between 0.2-1.8 and yellow to violet at pH between 7.0-8.8. In alkaline solutions, cresol red is present as a hydrophilic dianion. The dianion is small and is violet in color, possessing a larger conjugated system than the yellow hydroxyquinone.

Application

Cresol Red is a widely used pH indicator for routine pH assessments like microbial culture media preparations. In addition, Cresol red is a suitable constituent of PCR reagent systems. Cresol Red is used as a tracking dye in DNA, RNA (agarose), and protein (polyacrylamide) electrophoresis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atsushi Seki et al.
Analytica chimica acta, 582(1), 154-157 (2007-03-28)
Novel spectroscopic sensor based on a hetero-core structured fiber optic is described in this paper. The hetero-core structured fiber optic consists of multi mode fibers and a short piece of single mode fiber which was inserted in the multi mode
K Horie et al.
Analytical biochemistry, 137(1), 80-87 (1984-02-01)
A highly sensitive quantitative method was developed to detect protons released or taken up upon ligand binding. A small change in pH due to proton release or uptake was detected by measuring the difference in the absorbance of a pH
Paula C A Jerónimo et al.
The Analyst, 130(8), 1190-1197 (2005-07-16)
An optical biosensor based on immobilised carbonic anhydrase and its application to the determination of the anti-glaucoma agent acetazolamide by enzyme inhibition measurements, is described. The enzyme and a pH indicator dye, cresol red, were physically immobilised in overlapped sol-gel
K Elamrani et al.
Biochimica et biophysica acta, 727(1), 22-30 (1983-01-05)
The kinetics of H+/OH- diffusion across dimyristoyl phosphatidic acid bilayer membranes was measured by following the absorbance of the pH-sensitive indicator Cresol red (o-cresolsulfonphthalein) entrapped in single lamellar vesicles after rapidly changing the external pH in a stopped-flow apparatus. The
K Schilling et al.
Clinical chemistry, 39(2), 251-256 (1993-02-01)
If buffer/indicator systems are used as optical thermometers in multicuvettes, the temperature resolution is limited by errors of optical measurements produced mainly by variations of pathlength and blank transmittance of the wells. Theoretical and practical approaches show that, in multicuvettes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service