06913
4′-Aminobenzo-18-crown-6
technical
Synonym(s):
(Benzo-18-crown-6)-4′-ylamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H25NO6
CAS Number:
Molecular Weight:
327.37
Beilstein:
5296463
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical
Quality Level
impurities
≤10% water
storage temp.
2-8°C
SMILES string
Nc1ccc2OCCOCCOCCOCCOCCOc2c1
InChI
1S/C16H25NO6/c17-14-1-2-15-16(13-14)23-12-10-21-8-6-19-4-3-18-5-7-20-9-11-22-15/h1-2,13H,3-12,17H2
InChI key
PZXYILUXRGTFGD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
4′-Aminobenzo-18-crown-6 is a cyclic compound. It is widely used as an ionophore. It coordinates with metal ions more readily than other ligands because of its planar oxygen atoms which provide a strong negative potential barrier.
Application
4′-Aminobenzo-18-crown-6 can be easily functionalized with magnetic nanoparticles and used as a solid-phase extraction adsorbent for the determination of Pb2+. It can also be used as a metal complexing agent to prepare a variety of molecular complexes.
Other Notes
Derivative of 18-crown-6 which can be derivatized by alkylation or acylation
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
E. Ozeki et al.
Journal of the Chemical Society. Chemical Communications, 1353-1353 (1988)
S. Shinkai et al.
Journal of the American Chemical Society, 109, 4458-4458 (1987)
Bulletin of the Chemical Society of Japan, 61, 4385-4385 (1988)
R. Ungaro et al.
Journal of the American Chemical Society, 98, 5198-5198 (1976)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service