Skip to Content
Merck
All Photos(2)

Key Documents

06536

Supelco

trans-Cinnamaldehyde

analytical standard

Synonym(s):

trans-3-Phenyl-2-propenal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCHO
CAS Number:
Molecular Weight:
132.16
Beilstein:
1071571
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.6 (vs air)

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.622 (lit.)

bp

250-252 °C (lit.)

mp

−9-−4 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

[H]C(=O)\C=C\c1ccccc1

InChI

1S/C9H8O/c10-8-4-7-9-5-2-1-3-6-9/h1-8H/b7-4+

InChI key

KJPRLNWUNMBNBZ-QPJJXVBHSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Pharmacological Activity in Metabolic Diseases: Cinnamaldehyde is investigated for its potential to attenuate diabetic osteoporosis in a rat model, mediated through the modulation of the netrin-1/DCC-UNC5B signaling pathway. This suggests its applicability in therapeutic strategies against metabolic bone diseases (Ji et al., 2024).

  • Antibacterial and Biofilm Inhibition: The study reviews the inhibitory effects of natural compounds, including cinnamaldehyde, on the quorum sensing systems of Pseudomonas aeruginosa. This indicates its potential role in managing biofilm communities, which could be crucial for developing new antibacterial strategies (Shariati et al., 2024).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: cinnamomum

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wen-Xian Du et al.
Journal of agricultural and food chemistry, 60(32), 7799-7804 (2012-07-27)
The addition of plant essential oils to edible films and coatings has been shown to protect against bacterial pathogens and spoilage while also enhancing sensory properties of foods. This study evaluated the effect of adding 0.5 and 0.75% carvacrol (active
Marie E Barabas et al.
PloS one, 7(10), e47988-e47988 (2012-11-08)
Subpopulations of somatosensory neurons are characterized by functional properties and expression of receptor proteins and surface markers. CGRP expression and IB4-binding are commonly used to define peptidergic and non-peptidergic subpopulations. TRPA1 is a polymodal, plasma membrane ion channel that contributes
Mari Pau Balaguer et al.
International journal of food microbiology, 166(3), 369-377 (2013-09-14)
Gliadin films incorporating 1.5, 3 and 5% cinnamaldehyde (g/100g protein) were tested against food-spoilage fungi Penicillium expansum and Aspergillus niger in vitro, and were employed in an active food packaging system for sliced bread and cheese spread. Gliadin films incorporating
Yaoqi Tian et al.
Carbohydrate polymers, 91(2), 586-589 (2012-11-06)
In this study, the high-amylose corn starch-cinnamaldehyde inclusion complex was prepared by an ultrasound treatment and its releasing characteristic was investigated. The results showed that the ultrasound treatment (35°C, 10min and 250W) generated a higher encapsulation rate of 40.2% than
Katherine R Zodrow et al.
Langmuir : the ACS journal of surfaces and colloids, 28(39), 13993-13999 (2012-09-04)
Biofilm-associated infections are one of the leading causes of death in the United States. Although infections may be treated with antibiotics, the overuse of antibiotics has led to the spread of antibiotic resistance. Many natural antimicrobial compounds derived from edible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service