Skip to Content
Merck
All Photos(1)

Key Documents

04976

Supelco

4-Ring NOGE (Novolac glycidyl ether) mixture of isomers, chain-like or branched

analytical standard

Synonym(s):

4-Ring NOGE (Novolac glycidyl ether) mixture of isomers, chain-like or branched, NOGE 4-Ring (Novolac glycidyl ether) mixture of isomers, Novolac glycidyl ether 4-Ring

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H40O8
Molecular Weight:
636.73
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service