Skip to Content
Merck
All Photos(2)

Key Documents

03811

Supelco

α-HCH-d6

PESTANAL®, analytical standard

Synonym(s):

α-1,2,3,4,5,6-Hexachlorocyclohexane-d6, α-BHC-d6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D6Cl6
CAS Number:
Molecular Weight:
296.87
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

isotopic purity

≥99 atom % D

product line

PESTANAL®

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

mp

159-160 °C

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

[2H][C@]1(Cl)[C@]([2H])(Cl)[C@]([2H])(Cl)[C@@]([2H])(Cl)[C@@]([2H])(Cl)[C@]1([2H])Cl

InChI

1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4-,5+,6+/m1/s1/i1D,2D,3D,4D,5D,6D

InChI key

JLYXXMFPNIAWKQ-NXPQFDGTSA-N

Related Categories

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service