Skip to Content
Merck
All Photos(1)

Key Documents

03273

Supelco

Salicylaldehyde

analytical standard

Synonym(s):

2-Hydroxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CHO
CAS Number:
Molecular Weight:
122.12
Beilstein:
471388
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.2 (vs air)

vapor pressure

1 mmHg ( 33 °C)

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.573 (lit.)
n20/D 1.573

bp

197 °C (lit.)

mp

1-2 °C (lit.)

density

1.146 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

Oc1ccccc1C=O

InChI

1S/C7H6O2/c8-5-6-3-1-2-4-7(6)9/h1-5,9H

InChI key

SMQUZDBALVYZAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ziyad A Taha et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 81(1), 317-323 (2011-07-15)
Eight new lanthanide metal complexes [LnL(NO(3))(2)]NO(3) {Ln(III) = Nd, Dy, Sm, Pr, Gd, Tb, La and Er, L = bis-(salicyladehyde)-1,3-propylenediimine Schiff base ligand} were prepared. These complexes were characterized by elemental analysis, thermogravimetric analysis (TGA), molar conductivity measurements and spectral
Rhodium(III)-catalyzed dehydrogenative Heck reaction of salicylaldehydes.
Zhuangzhi Shi et al.
Angewandte Chemie (International ed. in English), 51(32), 8092-8096 (2012-06-30)
Mirta Rubčić et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(18), 5620-5631 (2012-03-27)
The Schiff base derived from salicylaldehyde and 2-amino-3-hydroxypyridine affords a diversity of solid forms, two polymorphic pairs of the enol-imino (D1 a and D1 b) and keto-amino (D2 a and D2 b) desmotropes. The isolated phases, identified by IR spectroscopy, X-ray crystallography, and (13)C
Wan Yen Lee et al.
Metallomics : integrated biometal science, 4(2), 188-196 (2011-12-03)
The copper(ii) complexes of two salicylaldehyde semicarbazones, HOC(6)H(4)CH[double bond, length as m-dash]N-NHCONR(2) [H(2)Bnz(2) (R = CH(2)Ph) and H(2)Bu(2) (R = Bu)], were evaluated for their DNA binding and cleavage properties by spectrophotometric DNA titration, ethidium bromide displacement assay and electrophoretic
Stephen K Murphy et al.
Organic letters, 13(23), 6216-6219 (2011-11-09)
A method has been developed for the intermolecular hydroacylation of homoallyl alcohols with salicylaldehydes to furnish homoaldol products in 50-98% yields. The method also applies to the hydroacylation of 2-hydroxystyrenes. This work highlights the use of hydroacylation as a unified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service