Skip to Content
Merck
All Photos(1)

Key Documents

02140

Sigma-Aldrich

Bis(2-ethylhexyl) adipate

≥97.0% (GC)

Synonym(s):

Adipic acid di(2-ethylhexyl) ester, DOA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CH2CO2CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
370.57
Beilstein:
1803774
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.447 (lit.)
n20/D 1.447

bp

175 °C/2 mmHg (lit.)

mp

<−70 °C (lit.)

density

0.925 g/mL at 20 °C (lit.)

functional group

ester

SMILES string

CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC

InChI

1S/C22H42O4/c1-5-9-13-19(7-3)17-25-21(23)15-11-12-16-22(24)26-18-20(8-4)14-10-6-2/h19-20H,5-18H2,1-4H3

InChI key

SAOKZLXYCUGLFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bis(2-ethylhexyl) adipate also known as dioctyl adipate (DOA) can be used as a plasticizer for improving the impact properties of polymers. DOA is also used to produce clear films for food packaging applications and in synthetic rubber industries due to its compatibility with nitrocellulose and ethylcellulose.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of bis (2-ethylhexyl) adipate in food products
Nerin C, et al.
Journal of Agricultural and Food Chemistry, 40(10), 1833-1835 (1992)
Use of plasticizers for electrochemical sensors
Recent Advances in Plasticizers (2012)
Polylactide (PLA) designed with desired end?use properties: 1. PLA compositions with low molecular weight ester?like plasticizers and related performances
Murariu M, et al.
Polymers For Advanced Technologies, 19(6), 636-646 (2008)
Julie Borch et al.
Reproductive toxicology (Elmsford, N.Y.), 19(4), 517-525 (2005-03-08)
This study aimed to characterize the effects of di(2-ethylhexyl) phthalate (DEHP) on the fetal rat testes and relate them to the effects seen in adults. Histopathological effects in fetal testes were examined with immunohistochemistry for anti-Mullerian hormone (AMH), 3beta-hydroxysteroid dehydrogenase
Stéphane C Fayoux et al.
Journal of food science, 72(4), E143-E154 (2007-11-13)
In order to better understand and optimize the sorption of limonin (the major navel orange juice bitter principle) by various plasticized polymeric films, a sorption and plasticizer migration study was carried out using high-performance liquid chromatography (HPLC) and gas chromatography

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service