Skip to Content
Merck
All Photos(1)

Key Documents

01240585

α-Hederin

primary reference standard

Synonym(s):

(3β,4α)-3-[[2-O-(6-Deoxy-α-L-mannopyranosyl)-α-L-arabinopyranosyl]oxy]-23-hydroxyolean-12-en-28-oic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H66O12
CAS Number:
Molecular Weight:
750.96
EC Number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@@H](O)CO[C@H]2O[C@H]3CC[C@@]4(C)[C@@H](CC[C@]5(C)[C@@H]4CC=C6[C@@H]7CC(C)(C)CC[C@@]7(CC[C@@]56C)C(O)=O)[C@]3(C)CO)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C41H66O12/c1-21-28(44)30(46)31(47)33(51-21)53-32-29(45)24(43)19-50-34(32)52-27-11-12-37(4)25(38(27,5)20-42)10-13-40(7)26(37)9-8-22-23-18-36(2,3)14-16-41(23,35(48)49)17-15-39(22,40)6/h8,21,23-34,42-47H,9-20H2,1-7H3,(H,48,49)/t21-,23-,24-,25+,26+,27-,28-,29-,30+,31+,32+,33-,34-,37-,38-,39+,40+,41-/m0/s1

InChI key

KEOITPILCOILGM-LLJOFIFVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H G Jeong et al.
Biochemistry and molecular biology international, 45(1), 163-170 (1998-06-23)
The protective effects of alpha-Hederin on carbon tetrachloride-induced hepatotoxicities were investigated in mice. Pretreatment with alpha-Hederin prior to the administration of carbon tetrachloride significantly prevented the increase in serum alanine aminotransferase (ALT) and lactate dehydrogenase (LDH) activity and lipid peroxidation
Marta Mendel et al.
Journal of ethnopharmacology, 146(1), 423-426 (2013-01-01)
The dry extract of Hedera helix leaves, due to its secretolytic and antispasmodic effects, is commonly used to produce pharmaceuticals applied in case of cough and other respiratory symptoms. The results of some in vitro studies as well as the
Bo Wang et al.
Organic & biomolecular chemistry, 10(44), 8822-8834 (2012-10-09)
We synthesized a diosgenyl saponin bearing a unique disaccharide from the natural product β-hederin, together with twelve glycosylated derivatives and determined their cytotoxicity against five different human cancer cell lines. Most of them showed weak cytotoxicity, with the exception of
A V Lekar' et al.
Bioorganicheskaia khimiia, 37(5), 679-684 (2012-02-16)
The molecular complexation of adenine and guanine with hederagenin 3-O-alpha-L-rhamnopyranosyl-(1-->2)-O-alpha-L-arabinopyranoside (alpha-hederin) and its 28-O-alpha-L-rhamnopyranosyl-(1-->4)-O-beta-D-glucopyranosyl-(1-->6)-O-beta-D-glucopyranosyl ester (hederasaponin C) was investigated for the first time using the method of electrospray ionization mass spectrometry. Guanine forms complexes more diverse in composition than adenine.
Geva Feller et al.
Planta medica, 76(16), 1847-1851 (2010-06-26)
α-Hederin, a natural triterpene saponin and its derivative kalopanaxsaponin I (ksI) exhibit cytotoxicity against various cancer cell lines and IN VIVO tumors. We studied the genetic variants contributing to the activity of these two anticancer compounds. Cell lines derived from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service