Skip to Content
Merck
All Photos(1)

Key Documents

01130595

Xanthohumol

primary reference standard

Synonym(s):

(E)-1-[2,4-Dihydroxy-6-methoxy-3-(3-methyl-2-butenyl)phenyl]-3-(4-hydroxyphenyl)propenone, 2′,4,4′-Trihydroxy-6′-methoxy-3′-prenylchalcone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22O5
CAS Number:
Molecular Weight:
354.40
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

SMILES string

COc1cc(O)c(C\C=C(/C)C)c(O)c1C(=O)\C=C\c2ccc(O)cc2

InChI

1S/C21H22O5/c1-13(2)4-10-16-18(24)12-19(26-3)20(21(16)25)17(23)11-7-14-5-8-15(22)9-6-14/h4-9,11-12,22,24-25H,10H2,1-3H3/b11-7+

InChI key

ORXQGKIUCDPEAJ-YRNVUSSQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference standard in the analysis of herbal medicinal products.

Other Notes

This compound is commonly found in plants of the genus: humulus

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Motyl et al.
Die Pharmazie, 67(1), 91-94 (2012-03-08)
Xanthohumol, the most abundant prenylated chalcone in hop (Humulus lupulus L.) cones, is well known to exert several promising pharmacological activities in vitro and in vivo. Among these, the chemopreventive, anti-inflammatory and anti-cancer effects are probably the most interesting. As
Roberto Benelli et al.
Biochemical pharmacology, 83(12), 1634-1642 (2012-03-27)
Although the vast majority of patients with acute lymphocytic leukemia (ALL) attain remission with modern therapies, relapsed leukemia will continue to be a common malignancy both in childhood and in adults, until new treatments are available. Therapeutic options for advanced
Marta Arczewska et al.
Biochimica et biophysica acta, 1828(2), 213-222 (2012-10-23)
Xanthohumol (XN) is the major prenylated flavonoid found in hop resin. It has attracted considerable attention in recent years due to its wide spectrum of biological activities and the beneficial effect on human health. Since lipid membrane is first target
Thomas E Ceremuga et al.
AANA journal, 81(3), 193-198 (2013-08-09)
The purpose of this study was to investigate the anxiolytic effects of xanthohumol, a component of Humulus lupulus (hops), and its potential interaction with the benzodiazepine binding site on the y-aminobutyric acid (GABAA) receptor in the male Sprague-Dawley rat. This
I Zajc et al.
Phytotherapy research : PTR, 26(11), 1709-1713 (2012-03-13)
Cytotoxicity and the mechanisms of cell death induced by xanthohumol (XN) were compared in normal and cancerous human cells as the differences may be relevant for the potential use of XN in cancer therapy. The cancer cells seemed to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service