Skip to Content
Merck
All Photos(1)

Key Documents

STREPWELLHB-RO

Roche

StreptaWell, High Bind (transparent, 12 x 8-well strips)

Synonym(s):

streptavidin-coated microplates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

packaging

pkg of 15 plates (11989685001 [96 wells])
pkg of 5 plates (11645692001 [12 x 8 well strips and frame])

Quality Level

manufacturer/tradename

Roche

storage temp.

2-8°C

General description

Transparent, streptavidin-coated microplates with high binding capacity:
  • Blocked volume: >300 μl
  • SA-coated area: 300 μl
  • Well geometry: C
Total biotin binding capacity: ≥20ng/well, ≥80pmol/well (competition assays; 300μL wells)
Total binding capacity: 1.5μg/well (biotin-labeled antibodies)

Application

StreptaWell can be used for colorimetric and fluorescent protein immunoassays.[1][2][3]

Features and Benefits

The unique coating process ensures homogenous and reproducible coatings with unique features:
  • Little variances between individual wells (intra-assay variance)
  • Little variances between different plates (inter-assay variance)
  • High lot-to-lot reproducibility
  • High signal-to-noise ratio
  • Low background
  • Almost no leaching

Quality

SA leaching (bleeding): <5ng/well

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

StreptaWell is a trademark of Roche

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean-Philippe Luzy et al.
Journal of biomolecular screening, 13(2), 112-119 (2008-01-25)
Adaptor proteins Grb7 and Grb2 have been implicated as being 2 potential therapeutic targets in several human cancers, especially those that overexpress ErbB2. These 2 proteins contain both a SH2 domain (Src homology 2) that binds to phosphorylated tyrosine residues
Gang-Ping Xue
Nucleic acids research, 30(15), e77-e77 (2002-07-26)
A rapid and quantitative DNA-binding assay was developed based on the translational fusion of a DNA-binding protein (DBP) with a Neocallimastix patriciarum beta-1,4-D-glucanase, CelD. CelD releases a fluorescent 4-methylumbelliferyl product from 4-methylumbelliferyl cellobioside. This hydrolysis activity was used to quantify
Ruth C Matthews et al.
Antimicrobial agents and chemotherapy, 47(7), 2208-2216 (2003-06-25)
Mycograb (NeuTec Pharma plc) is a human genetically recombinant antibody against fungal heat shock protein 90 (HSP90). Antibody to HSP90 is closely associated with recovery in patients with invasive candidiasis who are receiving amphotericin B (AMB). Using in vitro assays

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service