Skip to Content
Merck
All Photos(5)

Key Documents

PFABIKB

Roche

KAPA PROBE FAST

suitable for qPCR, 2 ×, ABI Prism®

Synonym(s):

probe qPCR, qPCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

Quality Level

shelf life

≤12 mo.

packaging

kit of 1 mL (100 x 20 μL rxn; KK4705)
kit of 10 mL (1000 x 20 μL rxn; KK4707)
kit of 5 mL (500 x 20 μL rxn; KK4706)
kit of 50 mL (5000 x 20 mL rxn; KK4714)
pkg of

manufacturer/tradename

Roche

concentration

2 ×

technique(s)

qPCR: suitable

compatibility

for use with Applied Biosystems® 5700
for use with Applied Biosystems® 7000
for use with Applied Biosystems® 7300
for use with Applied Biosystems® 7700
for use with Applied Biosystems® 7900HT
for use with StepOne
for use with StepOnePlus

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

KAPA PROBE FAST qPCR Master Mix (2X) ABI Prism® Kits provide fast and reproducible results for all probe-based qPCR applications. Kits contain a ready-to-use master mix for highly sensitive and accurate real-time PCR using sequence-specific fluorogenic probe chemistries, including hydrolysis probes (e.g. TaqMan®), FRET probes, and displacement probes (e.g. molecular beacons).

Application

KAPA PROBE FAST qPCR Master Mix (2X) ABI PRISM® is suitable for:
  • Gene expression analysis
  • Multiplex qPCR
  • SNP genotyping/allelic discrimination
  • Microarray validation.
  • Reverse transcription (RT)-PCR/RT-qPCR
  • Quantitative polymerase chain reaction (qPCR)
  • Genotyping

Features and Benefits

Accurately discriminate between alleles:
  • Exceptional differentiation of heterozygous and homozygous alleles
  • Discrete clusters in SNP genotyping assays

Achieve excellent reproducibility and efficiency:
  • Fast, high performance five-colored multiplex PCR
  • Similar abundance levels achieved with low copy number
  • Broad dynamic range of up to 10 orders of magnitude

Increase flexibility:
  • Compatible with both standard and fast cycling protocolsRetain accurate quantification

Quick Notes:
  • These kits are compatible with all fluorogenic probebased technologies, including hybridization probes (e.g., FRET), hydrolysis probes (e.g., TaqMan®) and displacement probes (e.g., molecular beacons).
  • Initial denaturation for 20 sec at 95ºC is sufficient for enzyme reactivation; however, optimal denaturation of complex targets may require up to 3 min denaturation.
  • For 2-step cycling, use 30 sec combined annealing/ extension/data acquisition.
  • For 3-step cycling, use 20 sec for primer annealing and 1 sec for extension/data acquisition.
  • Do not exceed 25 μL volumes.
  • Compatible with different qPCR protocols

Quality

KAPA PROBE FAST qPCR Master Mix (2X) ABI PRISM® is subjected to stringent quality control tests, is free of contaminating exo and endonuclease activity, and meets strict requirements with respect to DNA contamination levels.

Preparation Note

ROX/fluorescein reference dyes (depending on kit configuration) are light sensitive. Exposure to direct light for an extended period of time will result in loss of fluorescent signal intensity. Avoid repeated freezing and thawing. Always ensure that the product has been fully thawed and mixed before use.

KAPA PROBE FAST qPCR Master Mix (2X) ABI PRISM® is stable through 30 freeze-thaw cycles. Ensure that all reagents are stored protected from light at -20°C when not in use. When protected from light, reagents are stable in the dark at 4°C for at least one week, and may be stored at this temperature for short-term use provided that they do not become contaminated with microbes and/or nucleases.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Legal Information

ABI PRISM is a registered trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
ABI Prism is a registered trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
Applied Biosystems is a registered trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
StepOne is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
StepOnePlus is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
TaqMan is a registered trademark of Roche Molecular Systems, Inc.

Kit Components Only

Product No.
Description

  • qPCR Master Mix

  • ROX High incorporated in the reaction mix

  • KAPA Taq HotStart® DNA Polymerase

  • dNTPs

  • MgCl2 5 nM

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catecholamines facilitate VEGF-dependent angiogenesis via β2-adrenoceptor-induced Epac1 and PKA activation.
Garg J, et al.
Oncotarget, 8(27), 44732-44732 (2017)
Heme oxygenase-1 induction by rosiglitazone via PKCa/AMPKa/p38 MAPKa/SIRT1/PPAR? pathway suppresses lipopolysaccharide-mediated pulmonary inflammation.
C Rou-Ling, et al.
Biochemical Pharmacology, 148, 222-237 (2018)
Tumor necrosis factor-alpha induces VCAM-1-mediated inflammation via c-Src-dependent transactivation of EGF receptors in human cardiac fibroblasts.
L Chih-Chung, et al.
Journal of Biomedical Science, 22(1), 53-53 (2015)
Jonathan B Wagner et al.
Journal of clinical pharmacology, 58(6), 823-833 (2018-02-23)
This study investigated the impact of allelic variation in SLCO1B1, a gene encoding for the liver-specific solute carrier organic anion transporter family member 1B1 protein (SLCO1B1), on simvastatin and simvastatin acid (SVA) systemic exposure in children and adolescents. Participants (8-20
Laura Raiko et al.
Experimental dermatology, 21(8), 586-591 (2012-05-30)
Mutations in the ATP2C1 gene encoding Ca(2+) /Mn(2+) ATPase SPCA1 cause Hailey-Hailey disease (HHD, OMIM 16960). HHD is characterized by epidermal acantholysis. We attempted to model HHD using normal keratinocytes, in which the SPCA1 mRNA was down-regulated with the

Articles

An overview of directed evolution and the methods for generating proteins with optimized or entirely new functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service