EHIFIP-RO
Roche
Expand™ High FidelityPLUS PCR System
sufficient for ≤500 reactions (03300226001), sufficient for ≤2,500 reactions (03300234001), suitable for PCR, High Fidelity PCR, hotstart: no, dNTPs included: no
Synonym(s):
high fidelity pcr plus system, expand
About This Item
Recommended Products
usage
sufficient for ≤2,500 reactions (03300234001)
sufficient for 2500 reactions
sufficient for ≤500 reactions (03300226001)
sufficient for 500 reactions
Quality Level
feature
High Fidelity PCR
dNTPs included: no
hotstart: no
packaging
pkg of 2,500 U (03300234001 [10x250 U])
pkg of 500 U (03300226001 [2x250 U])
manufacturer/tradename
Roche
parameter
72 °C optimum reaction temp.
technique(s)
PCR: suitable
input
purified DNA
storage temp.
−20°C
Related Categories
General description
The Expand High FidelityPLUS PCR System is designed to amplify fragments up to 5 kb from all types of DNA with high yield and fidelity. It amplifies DNA with sixfold greater fidelity than Taq DNA polymerase alone.
Expand High FidelityPLUS PCR System also incorporates dUTP. Cross-contamination can therefore be prevented by a pretreatment of dUTP containing DNA with heat-labile Uracil-DNA Glycosylase before amplification. It is suited for the simultaneous amplification of a large number of targets.
Application
Use Expand High FidelityPLUS PCR System for:
- PCR
- RT-PCR
- Primer extension
- Carryover prevention
Features and Benefits
- Safeguard against cross-contamination.
- Obtain sixfold higher fidelity and higher yields,
- Efficiently label DNA.
Packaging
Quality
Unit Definition
Other Notes
Legal Information
Kit Components Only
- Expand High FidelityPLUS Enzyme Blend 5 U/μl
- Expand High FidelityPLUS Reaction Buffer, with 7.5 mM MgCl2 5x concentrated
- Expand High FidelityPLUS Reaction Buffer, without MgCl2 5x concentrated
- MgCl2 Stock Solution 25 mM
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service