Skip to Content
Merck
All Photos(5)

Key Documents

2GRHSKB

Roche

KAPA2G Robust HotStart PCR Kit

Synonym(s):

PCR kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

usage

sufficient for 1000 reactions
sufficient for 200 reactions
sufficient for 500 reactions
sufficient for 5000 reactions

Quality Level

shelf life

≤12 mo.

feature

Difficult Templates/Specialty Enzymes PCR
dNTPs included: no
hotstart

packaging

pkg of 100 U (KK5522)
pkg of 250 U (KK5515)
pkg of 2500 U (KK5525)
pkg of 500 U (KK5517)

manufacturer/tradename

Roche

technique(s)

PCR: suitable

input

crude DNA

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The second-generation KAPA2G Robust DNA Polymerase was evolved to solve inconsistent amplification across a broad range of amplicon types (GC- and AT-rich). KAPA2G Robust HotStart PCR Kit enables higher processivity and specific activity, which translates to robust PCR performance, high sensitivity, and improved tolerance to common inhibitors. The high performance of the KAPA2G Robust DNA Polymerase is ideally suited for challenging PCR applications and difficult samples, eliminating the need for optimization using multiple enzymes and protocols.

Application

KAPA2G Robust HotStart® PCR Kit has been used in:
  • PCR amplification of saliva samples
  • PCR amplification for direct sequencing
  • Nested PCR
  • Amplification of GC- or AT-rich templates
  • Templates containing common PCR inhibitors e.g. salts, urea, SDS, or ethanol
  • Crude sample PCR e.g. buccal swabs, cultured mammalian, yeast, or bacterial cells
  • Colony PCR
  • RT-PCR

Biochem/physiol Actions

KAPA2G Robust HotStart® DNA Polymerase generated DNA are similar to fragments generated with wild-type Taq DNA polymerase. The fidelity of KAPA2G Robust HotStart is similar to that of wild-type Taq; it has an error rate of approximately 1 error per 1.7 x 105 nucleotides incorporated. It may be used in restriction enzyme digestion, cloning, and sequencing. Like wild-type Taq, KAPA2G Robust HotStart has 5′→3′ polymerase and 5′→3′ exonuclease activities, but no 3′→5′ exonuclease (proofreading) activity.

Features and Benefits

Efficient amplification of GC- and AT-rich targets
  • Robust performance across a wide range of GC- and AT-rich templates
  • Increased PCR success rates

Improved tolerance to common PCR inhibitors
  • Efficient amplification from crude samples
  • Higher yield and sensitivity per unit of enzyme

Unrivalled performance in colony PCR
  • Higher yields and improved consistency of PCR direct from E. coli and yeast cells

Quick Notes:
  • KAPA2G Robust HotStart PCR Kits contain KAPA2G Robust HotStart DNA Polymerase, engineered for high processivity and inhibitor tolerance.
  • Both purified genomic DNA and crude samples (e.g. colony PCR) can be used as template.
  • Use 0.5 U of KAPA2G Robust HotStart DNA Polymerase per 25 μL reaction. More challenging PCRs (GCrich,crude sample) may require higher enzyme concentrations.
  • Use 15 sec/kb extension time per cycle, and increase to 30-60 sec/kb for difficult amplicons or templates.
  • KAPA2G Buffers contain 1.5 mM MgCl2 at 1X.
  • Additional MgCl2 may be added using the 25 mM MgCl2 solution provided in the kit.
  • KAPA2G Buffer A is optimized for high yield, specificity, and sensitivity.
  • KAPA2G Buffer B is recommended for inhibitorcontaminated template samples.
  • KAPA2G GC Buffer is recommended for GC-rich PCR (>70% GC).
  • KAPA Enhancer 1 can be used with KAPA2G Buffer A or B, particularly for GC-rich assays.
  • Reaction products are 3′-dA-tailed and may be cloned into TA cloning vectors.

Quality

Each batch of KAPA2G Robust HotStart DNA Polymerase is confirmed to contain <2% contaminating protein (Agilent Protein 230 Assay). KAPA2G Robust HotStart PCR Kits are subjected to stringent quality control tests, are free of contaminating exo- and endonuclease activity, and meet strict requirements with respect to DNA contamination levels.

Preparation Note

Always ensure that the product has been fully thawed and mixed before use. Reagents may be stored at 4°C for short-term use (up to 1 month). Return to -20°C for long-term storage. Provided that all components have been handled carefully and not contaminated, the kit is not expected to be compromised if left (unintentionally) at room temperature for a short period of time (up to 3 days). Long-term storage at room temperature and 4°C is not recommended. Please note that reagents stored at temperatures above -20°C are more prone to degradation when contaminated during use, and therefore storage at such temperatures is at the user′s own risk.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Legal Information

HOTSTART is a registered trademark of Molecular BioProducts, Inc.

Kit Components Only

Product No.
Description

  • KAPA2G Robust Standard or HotStart® DNA Polymerase 5 U/μL

  • 5X KAPA2G Buffer A

  • 5X KAPA2G Buffer B

  • 5X KAPA2G GC Buffer with MgCl2

  • 5X KAPA Enhancer 1

  • MgCl2 25 mM

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - STOT SE 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bette K Kleinschmidt-DeMasters et al.
The American journal of surgical pathology, 39(4), 528-540 (2015-01-13)
Epithelioid glioblastomas (E-GBMs) manifest BRAF V600E mutation in up to 50% of cases, compared with a small percentage of ordinary GBMs, suggesting that they are best considered variants rather than a different pattern of GBM. Availability of a targeted therapy
Zhiqiang Li et al.
Cells, 9(4) (2020-04-15)
Mast cells have been shown to affect the control of infections with the protozoan parasite Giardia intestinalis. Recently, we demonstrated that Giardia excretory-secretory proteins inhibited the activity of the connective tissue mast cell-specific protease chymase. To study the potential role
Colin G Deyoung et al.
Journal of research in personality, 45(4), 364-371 (2011-08-02)
The personality trait Openness/Intellect reflects the tendency to be imaginative, curious, perceptive, artistic, and intellectual-all characteristics that involve cognitive exploration. Little is known about the biological basis of Openness/Intellect, but the trait has been linked to cognitive functions of prefrontal
Use of surface enhanced blocking (SEB) electrodes for microbial cell lysis in flow-through devices.
Talebpour A, et al.
PLoS ONE, 9(7), e102707-e102707 (2014)
Novel TP53 gene mutation and correlation with p53 immunohistochemistry in a mixed epithelial carcinoma of the endometrium.
Sholl A B, et al.
Gynecologic oncology case reports, 3, 11-11 (2013)

Articles

An overview of directed evolution and the methods for generating proteins with optimized or entirely new functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service