Skip to Content
Merck
All Photos(1)

Key Documents

11815016001

Roche

Anti-HA Affinity Matrix

from rat IgG1

Synonym(s):

affinity matrix, anti-ha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rat

Quality Level

conjugate

agarose conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F10, monoclonal

form

slurry

packaging

pkg of 1 mL (settled resin volume)

manufacturer/tradename

Roche

isotype

IgG1

epitope sequence

YPYDVPDYA

capacity

2-8 nmol/mL binding capacity

storage temp.

2-8°C

Related Categories

General description

Anti-HA High Affinity antibody (clone 3F10) recognizes the 9-amino acid sequence YPYDVPDYA, derived from the human influenza hemagglutinin (HA) protein. This epitope is also recognized in fusion proteins that are derived from mammalian, bacterial, and yeast expression vectors, regardless of its position (N-terminal, C-terminal or internal)

Specificity

Anti-HA High Affinity antibody (clone 3F10) recognizes the 9-amino acid sequence YPYDVPDYA, derived from the human influenza hemagglutinin (HA) protein. This epitope is also recognized in fusion proteins that are derived from mammalian, bacterial, and yeast expression vectors, regardless of its position (N-terminal, C-terminal or internal).

Immunogen

Amino acids 98-106 from the human influenza virus hemagglutinin protein

Application

Following immunoprecipitation or purification, the tagged protein of interest may be analyzed by:
  • Western blotting using the Anti-HA antibody
  • Silver staining (or similar protein stain)
  • In coimmunoprecipitation.
Use Anti-HA Affinity Matrix for:
  • Immunoprecipitation of HA-tagged proteins from mammalian, bacterial, and yeast cell extracts
  • Affinity column purification of HA-tagged proteins from crude protein extracts

Features and Benefits

  • Achieve enhanced specificity and reduced cross-reactivity of Anti-HA High Affinity.
  • Use nondenaturing conditions for elution of your HA-tagged protein of choice.
  • Purify even rarely expressed HA-tagged proteins.
  • Highly specific to YPYDVPDYA.
  • Suitable for purification of proteins containing HA-epitop as N-terminal, C-terminal or internal fusion.
  • Applicable with crued cell extracts from mammalian, bacterial and yeast expression systems.

Quality

Each lot of Anti-HA Affinity Matrix is tested for its ability to purify a HA-tagged protein expressed in stably transfected cells from a crude mammalian extract. The immunoprecipitate is analyzed by western blot using Anti-HA, High Affinity antibody.

Physical form

1 ml settled resin of Anti-HA Affinity Matrix in PBS containing 0.09% sodium azide (w/v); 2 ml suspension equals to 1 ml bed volume. One plastic column with top and bottom caps is included.

Analysis Note

Yield of 9 nmol purified protein/ml affinity matrix was determined using a whole-cell bacterial extract containing expressed HA-tagged bacterial alkaline phosphatase.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geetha Hewawasam et al.
Molecular cell, 40(3), 444-454 (2010-11-13)
Cse4 is a variant of histone H3 that is incorporated into a single nucleosome at each centromere in budding yeast. We have discovered an E3 ubiquitin ligase, called Psh1, which controls the cellular level of Cse4 via ubiquitylation and proteolysis.
Abdirahman Abdi et al.
Cellular and molecular life sciences : CMLS, 67(19), 3355-3369 (2010-06-29)
Over the last decade, several protein kinases inhibitors have reached the market for cancer chemotherapy. The kinomes of pathogens represent potentially attractive targets in infectious diseases. The functions of the majority of protein kinases of Plasmodium falciparum, the parasitic protist
Wim Schepers et al.
The Journal of biological chemistry, 287(53), 44130-44142 (2012-11-17)
The readdition of an essential nutrient to starved, fermenting cells of the yeast Saccharomyces cerevisiae triggers rapid activation of the protein kinase A (PKA) pathway. Trehalase is activated 5-10-fold within minutes and has been used as a convenient reporter for
Stephen C Kales et al.
PloS one, 9(1), e87116-e87116 (2014-01-28)
The Cbl proteins (Cbl, Cbl-b, and Cbl-c) are a highly conserved family of RING finger ubiquitin ligases (E3s) that function as negative regulators of tyrosine kinases in a wide variety of signal transduction pathways. In this study, we identify a
Jinyan Cheng et al.
Journal of virology, 94(7) (2020-01-17)
Stress granules (SGs) are formed in the cytoplasm under environmental stress, including viral infection. Human enterovirus D68 (EV-D68) is a highly pathogenic virus which can cause serious respiratory and neurological diseases. At present, there is no effective drug or vaccine

Protocols

To concentrate the protein eluate, Roche recommends following the immunoprecipitation protocol given in the package insert from step 2 to 5. Use 50 μl of resuspended Anti-HA Affinity Matrix in a microcentrifuge tube.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service