Skip to Content
Merck
All Photos(1)

Key Documents

11051440001

Roche

dATP

=98%, solution, 100 mM, suitable for DNA sequencing, suitable for DNA amplification

Synonym(s):

2′-Deoxyadenosine 5′-triphosphate sodium salt solution, 2′-deoxy-adenosine-5′-triphosphate, deoxy-adenosine-5′-triphosphate, 2′-, dATP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116100

Quality Level

description

Lithium salt

Assay

98%

form

solution

usage

sufficient for 6,250 standard reactions (PCR)

packaging

pkg of 250 μL (25 μmol; 100 mM)

manufacturer/tradename

Roche

concentration

100 mM

technique(s)

DNA amplification: suitable
DNA sequencing: suitable
DNA synthesis: suitable
PCR: suitable

storage temp.

−20°C

SMILES string

[Na+].Nc1ncnc2n(cnc12)[C@H]3C[C@H](O)[C@@H](COP(O)(=O)OP(O)(=O)OP(O)([O-])=O)O3

InChI

1S/C10H16N5O12P3.Na/c11-9-8-10(13-3-12-9)15(4-14-8)7-1-5(16)6(25-7)2-24-29(20,21)27-30(22,23)26-28(17,18)19;/h3-7,16H,1-2H2,(H,20,21)(H,22,23)(H2,11,12,13)(H2,17,18,19);/q;+1/p-1/t5-,6+,7+;/m0./s1

InChI key

YJWCICGGRLOGEH-VWZUFWLJSA-M

Looking for similar products? Visit Product Comparison Guide

General description

2′-Deoxyadenosine 5′-triphosphate (dATP) is made up of a nucleobase attached to deoxyribose and a 5′-hydroxyl on the sugar bound to a chain of three phosphate residues. dATP is used by cells for synthesis of DNA by DNA polymerases.
dATP is a 100 mM clear, colorless solution of the lithium salt (pH 7).Download the CofA, SDS and IFU s

Application

2′-Deoxyadenosine 5′-triphosphate sodium salt solution is used in DNA sysnthesis reactions such as PCR, DNA sequencing and molecular cloning techniques.
Use dATP, lithium salt, for DNA sequencing, labeling, and all types of amplification reactions or primer-extension reactions.
dATP has been used to resume transcription.

Biochem/physiol Actions

Deoxyadenosine-5-triphosphate (dATP) plays a major role in the initiation of apoptosis in vitro. dATP is considered as an inhibitor of mammalian ribonucleotide reductase, thereby DNA synthesis.

Quality

Typical analysis:
  • ≤98% dATP (HPLC), ≤1.5% dADP (HPLC), ≤0.5% dAMP (HPLC)
  • Absorbance determination A250/A260, A280/A260, A290/A260
  • DNase activity negative, according to the current Quality Control procedures
  • Suitability for PCR

Unit Definition

1 μmol dATP = 15.0 A260 units

Other Notes

For life science research only. Not for use in diagnostic procedures.
Our PCR Grade deoxynucleotides are even purer than the lithium salts, to ensure excellent performance in your PCRs. PCR Grade deoxynucleotides contain the sodium salt of the nucleotide in PCR Grade Water.
For your convenience, some of our deoxynucleotides are also available in:
  • nucleotide sets (all 4 nucleotides in separate vials)
  • ready-to-use nucleotide mixes (premixed solutions of all 4 nucleotides)
They are also vital components of:
  • kits (dNTPs, buffer, and polymerase in separate vials)
  • ready-to-use mixes [polymerase, nucleotides, and buffer (premixed 2x solutions)]

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mfd Dynamically Regulates Transcription via a Release and Catch-Up Mechanism
Le TT, et al.
Cell (2018)
Stable suppression of the R2 subunit of ribonucleotide reductase by R2-targeted short interference RNA sensitizes p53(-/-) HCT-116 colon cancer cells to DNA-damaging agents and ribonucleotide reductase inhibitors
Lin ZP, et al.
The Journal of Biological Chemistry, 279(26), 27030-27038 (2004)
Induction of apoptotic program in cell-free extracts: requirement for dATP and cytochrome c
Liu X, et al.
Cell, 86(1), 147-157 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service