Skip to Content
Merck
All Photos(1)

Key Documents

10800058001

Sigma-Aldrich

Storage Box

capacity 50 wells, pkg of 1 ea, for tubes, 0.5 ‑ 2 mL

Synonym(s):

enzyme storage box, freezer storage box

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122804

feature

for tubes 0.5 ‑ 2 mL

packaging

pkg of 1 ea

manufacturer/tradename

Roche

capacity

50 wells

shipped in

ambient

Related Categories

General description

Box with removable styrofoam insert (0.5 to 2.0 ml capacity) for the storage of 50 microcentrifuge tubes.
Temperature-resistant to -70 °C.

Application

Storage box is used for the storage of 0.5-2mL microfuge tubes.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morgan H McCoy et al.
Journal of visualized experiments : JoVE, 9(89)(89) (2014-07-22)
It is crucial to identify cytomegalovirus (CMV) infection in the gastrointestinal (GI) tract of immunosuppressed patients, given their greater risk for developing severe infection. Many laboratory methods for the detection of CMV infection have been developed, including serology, viral culture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service