10102121001
Roche
Adenosine Deaminase (ADA)
from calf intestine
Synonym(s):
ADA, Adenosine Deaminase
About This Item
Recommended Products
biological source
bovine intestine
Quality Level
form
suspension
specific activity
~200 units/mg protein (At 25 °C with adenosine as the substrate.)
packaging
pkg of 1 mL (10 mg)
manufacturer/tradename
Roche
technique(s)
activity assay: suitable
color
white
optimum pH
7.0-7.4
solubility
water: miscible
suitability
suitable for chemiluminescence
NCBI accession no.
UniProt accession no.
application(s)
life science and biopharma
foreign activity
AMP-deaminase <0.01%
AP <0.01% (with pre incubation)
Guanase <0.01%
NP <0.01%
storage temp.
2-8°C
Gene Information
bovine ... ADA(280712)
Related Categories
General description
Adenosine deaminase (ADA) is a zinc metalloenzyme.
Application
Adenosine deaminase (ADA) has been used in lipolysis assay along with other reagents.
Biochem/physiol Actions
Quality
Physical form
Other Notes
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
No data available
Flash Point(C)
No data available
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service