ZLXL51040
Milli-Q® HX Water Purification System
Centralized pure water solution for up to 800 L/dayType 2 water; for low chlorine feed water.
About This Item
AC/DC input
100 - 230 V (For region specific voltage information, see footnote below)
Quality Level
product line
Milli-Q® HX
packaging
pkg of 1 unit
parameter
4 L/min flow rate
40 L/hr make-up flow rate
78 kg operating weight (172 lb)
800 L/day max. usage
resistivity
>5 MΩ-cm, 25°C
H × W × D
124 cm (48.8 in.) × 54.3 cm (21.4 in.) × 54.2 cm (21.3 in.)
total organic carbon (TOC) residue
<30 ppb (Product water)
input
feed water nature potable tap water
output
product water quality: type 2 water (> 5 MΩ·cm)
conductivity
<0.2 μS/cm at 25 °C
Looking for similar products? Visit Product Comparison Guide
General description
- Milli-Q® HX 7000 systems are designed to deliver high and consistent pure water quality for regular laboratory applications and for instrument feed.
- The Milli-Q® HX 7040 high-flow system is designed to produce up to 800 L/day of Type 2 pure water from potable tap feed water, making them ideal for large laboratories or lab buildings.
- Tap water is first treated by the Progard® pretreatment cartridge to efficiently remove particles, colloids, free chlorine and hardness. Advanced reverse osmosis (RO) purification then removes 95-99% of ions, and 99% of all dissolved organics (>200 Da), microorganisms and particles. Patented E.R.A.® (Evolutive Reject Adjustment) technology automatically assesses feed water quality to produce a constant flow rate and optimize water recovery (between 45% and 75%). The Elix® electrodeionization (EDI) module further removes ions to typically reach resistivity >5 MΩ.cm (at 25 °C).
- Processed pure water is then irradiated by a bactericidal UV lamp before entering a storage tank or SDS 500 storage, protection and distribution system. The SDS 500 reservoir maintains the consistent purity of stored water. Its vent filter provides effective protection against airborne contaminants and the Automatic Sanitization Module (ASM) prevents bacterial growth and biofilm formation.
- Milli-Q® HX 7040 product water can then be sent by a distribution pump to feed instruments or a distribution loop. These can be integrated into a centralized system, providing total control of all parameters within the system itself as well as within the external pure water distribution loop.
- The system′s large interactive touchscreen display offers a superior communication interface that shows main data (water production status, storage level, dispensing status), consumables exhaustion status, alerts status, and handy wizards to guide users through maintenance procedures.
- These central systems are embedded with our unique MyMilli-Q™ RemoteCare online monitoring and service capability, which operates as an extension of Milli-Q® Services. This digital service lets you securely and remotelymanage your system, supporting your lab′s optimal productivity.
Application
- Type 2 Elix® product water from Milli-Q® HX 7000 systems is suitable for microbiological media preparation, buffer preparation, and in manufacturing of chemical and biochemical reagents.
- It may be used to feed equipment such as weathering and stability test chambers, glassware washers, autoclaves and hydrogen generators. It may also feed a Milli-Q® ultrapure water (Type 1) system.
- Because pure analytical-grade water is required for a broad range of laboratory applications, Milli-Q® HX 7000 systems are a perfect fit for labs everywhere, including those in the pharmaceutical, clinical, chemical, metallurgical, cosmetics, food and beverage, electronics and biotech sectors.
Features and Benefits
- Progard® pretreatment packs efficiently remove particles (0.5 μm filter), free chlorine and colloids from tap water.
- Advanced RO and E.R.A.® technologies optimize water recovery to maintain constant flow rate and decrease water consumption. Users save water, time and money.
- The Elix® EDI module ensures constant quality of pure water and low running costs.
- UV lamps and final filtration provide full bacterial control.
Components
- The indicated price currently corresponds to the above catalogue number only; it is subject to change at any time without notice.
- This catalogue number does not include consumables, accessories and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
- Because our customers each have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you, do not hesitate to contact us!
Other Notes
- Organism Retention: Microorganisms
- Mode of Action: Filtering and UV
- Application: General laboratory analysis
- Intended Use: Water purification
- Instructions for Use: This item provides water filtered through 0.22 μm sterilizing-grade membranes and exposed to bactericidal UV lamp. Refer to system equipment user guide section "Using the System" (available on USB key in the system box)
- Storage Statement: Store in dry location
- Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.
Footnote
- 230 V systems are required for Europe, Africa and Middle East regions, and certain LATAM and APAC countries
- 120 V systems are required for North America, Taiwan, and certain LATAM countries
- 110 V systems are required for Japan
Legal Information
Disclaimer

Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service