Skip to Content
Merck
All Photos(1)

Key Documents

UC0E03

Sigma-Aldrich

UCOE® Dual Expression Puromycin Vector Set

Synonym(s):

UCOE Mammalian Protein Expression Vectors, UCOE Mammalian Protein Expression Plasmids, Ubiquitous Chromatin Opening Element, UCOE03

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106621
eCl@ss:
34360190
NACRES:
NA.51

shipped in

dry ice

Quality Level

General description

The Ubiquitous Chromatin Opening Element (UCOE) technology offers significant advances in recombinant mammalian protein expression. Unlike traditional vectors, UCOE-containing vectors have the capacity to ease the process of isolating cell line clones that express high-levels of recombinant proteins. UCOE sequences promote reproducible and stable high level expression of a linked gene by altering chromatin structure to a transcriptionally permissive state that negates epigenetic-mediated (e.g. DNA methylation) silencing.

CET 1019 HD-puro-SceI is a mammalian expression plasmid vector containing both the mouse Rps3 UCOE and the human HNRPA2B1-CBX3 UCOE each upstream of a strong human cytomegalovirus (hCMV) immediate early promoter-enhancer element.

CET 1019 AD-puro-SceI is a mammalian expression plasmid vector containing both the mouse Rps3 UCOE and the human HNRPA2B1-CBX3 UCOE each upstream of a guinea pig CMV (gpCMV) promoter-enhancer element.

The CET 1019 HD-puro-SceI and CET 1019 AD-puro-SceI vectors possess the following features:

  • A multiple cloning site downstream of the UCOE-CMV combinations containing unique restriction enzyme sites to permit easy insertion of any gene of interest (cDNA or genomic)
  • A polyadenylation element from the SV40 early region located downstream of the multiple cloning site for appropriate mRNA 3 end formation
  • A puromycin antibiotic resistance gene for selection of stably transfected mammalian cells
  • A β-lactamase gene for plasmid selection and propagation in transformed prokaryotic cells in the presence of ampicillin
  • An I-SceI homing restriction endonuclease site for linearization of constructs prior to transfection into mammalian cells, which favors integration into the target cell genome via the pre-generated free DNA ends and thus retains the integrity of the UCOE-CMV-transgene cassettes


THIS PRODUCT IS ONLY AVAILABLE FOR SALE TO ACADEMIC INSTITUTIONS OR NOT-FOR-PROFIT ENTITIES FOR USE UNDER THIS PRODUCT LABEL LICENSE. FOR INFORMATION ON COMMERCIAL LICENSING OF THE PATENTED UCOE TECHNOLOGY, PLEASE CONTACT licensing@emdmillipore.com.

Components

1) 10ug CET 1019 HD-puro-SceI (+) Vector (CS221292)

2) 10ug CET 1019 AD-puro-SceI (+) Vector (CS221293))

3) 10ug DC HD-puro (-) Vector (CS221283)

4) 10ug DC AD-puro (-) Vector (CS221295)

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

UCOE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service