Skip to Content
Merck
All Photos(1)

Key Documents

TMS-002

Sigma-Aldrich

EmbryoMax® L-Glutamine Solution (100X), 200mM

The EmbryoMax L-Glutamine Solution (100X), 200mM is available in a 100 mL format and may be used for routine mouse embryonic stem cell culture applications.

Synonym(s):

L-Glutamine solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352205
eCl@ss:
32160801
NACRES:
NA.75
Pricing and availability is not currently available.

Quality Level

form

liquid

manufacturer/tradename

Specialty Media
EmbryoMax®

technique(s)

cell culture | stem cell: suitable

input

sample type: mouse embryonic stem cell(s)
sample type induced pluripotent stem cell(s)

General description

EmbryoMax® L-Glutamine Solution (100X), 200mM may be used for routine mouse embryonic stem cell culture applications.

Application

EmbryoMax® L-Glutamine Solution (100X), 200mM has been used as a component of the high glucose Dulbecco′s modified Eagle medium (DMEM) to culture human hepatic stellate cell line[1] and mouse hippocampal neuronal cell line[2]. It has also been used as a supplement in Roswell Park Memorial Institute (RPMI) media for the maintenance of mouse dendritic cells.[3]

Storage and Stability

Store at -20°C for 1 year.

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minhui Jiang et al.
Neuroscience, 479, 48-59 (2021-10-15)
Diagnosis of major depressive disorder (MDD) is perplexing due to its multifactorial etiologies. Here, we isolated exosomes from the peripheral blood of MDD patients and healthy control subjects for mass spectrometry-based label-free quantitative proteomics. We identified that SERPINF1 is significantly
Huan Liu et al.
Biochemical and biophysical research communications, 514(3), 913-918 (2019-05-16)
Liver fibrosis is a common consequence of various chronic hepatitis and liver injuries. The myofibroblasts, through the accumulation of extracellular matrix (ECM) proteins, are closely associated with the progression of liver fibrosis. However, the molecular mechanisms underlying transcriptional regulation of
Ernest T Chivero et al.
Frontiers in cell and developmental biology, 8, 573-573 (2020-08-28)
MicroRNA-124 (miR-124), a brain-enriched microRNA, is known to regulate microglial quiescence. Psychostimulants such as cocaine have been shown to activate microglia by downregulating miR-124, leading, in turn, to neuroinflammation. We thus rationalized that restoring the levels of miR-124 could function

Protocols

Step-by-step protocol for ADCC assays using cryopreserved NK effector cells including tips and tricks, assay development, and data analysis methods.

Discover our primary human hepatocytes and plating and maintenance media formulations. Explore our protocol for plateable primary hepatocyte cell culture.

Discover our primary human hepatocytes and plating and maintenance media formulations. Explore our protocol for plateable primary hepatocyte cell culture.

Discover our primary human hepatocytes and plating and maintenance media formulations. Explore our protocol for plateable primary hepatocyte cell culture.

See All

Related Content

Explore our protocol for suspension primary hepatocyte cell culture. Discover our primary human hepatocytes and plating and maintenance media formulations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service