SCT039
BioTracker Green H2O2 Live Cell Dye
Live cell imaging probe for detection of hydrogen peroxide (H<sub>2</sub>O<sub>2</sub>) in live cells.
Synonym(s):
BioTracker H₂O₂ Dye, Green Cell Dye
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.47
Recommended Products
form
lyophilized
application(s)
cell analysis
detection method
fluorometric
Related Categories
General description
Hydrogen peroxide (H2O2) is involved in therapeutic processes such as wound healing, anti-bacterial defense, stem cell proliferation, and an adaptive response in astrocytes that leads to neuronal protection. However, over-production of H2O2 exerts toxic effects on the cell and its surrounding environment. Over-production of H2O2 is connected to serious pathological conditions such as cancer, ageing, diabetes, and neurodegenerative diseases.BioTracker Green H2O2 live cell dye is a fluorescent probe that fluoresces upon reaction with H2O2 , but does not react with other ROS such as hydroxyradical (-OH), superoxide (O2-), hypochlorous acid (HOCl), singlet oxygen (1O2), and nitric oxide (NO). This dye is cell permeable and initially has low reactivity with H2O2 but it is quickly hydrolyzed by intracellular esterases to generate a reactive and cell impermeable product that fluoresces upon reaction with H2O2. Spectral PropertiesAbsorbance (max): 492 nmEmission (max): 516 nm (Green)Research Categories:InflammationLive Cell Dye
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
604.2 °F
Flash Point(C)
317.9 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service