SCC429
ANBL-6 Plasma Cell Myeloma Cell Line
Synonym(s):
ANBL6
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.81
packaging
vial of ≥1X10⁶ cells/vial mg
Quality Level
manufacturer/tradename
Millipore
growth mode
N/A
technique(s)
cell culture | stem cell: suitable
shipped in
liquid nitrogen
storage temp.
−196°C
General description
ANBL-6 is a well-established model for MM.3 In culture ANBL-6 cell proliferation is dependent on IL-6, a critical component of the bone marrow microenvironment supporting tumor growth.2 Transcriptomic analysis of ANBL-6 cells demonstrated a high correlation with genetic signatures of primary MM tumors.2 ANBL-6 cells home to hematopoietic tissues in a mouse model, with a growth phenotype characteristic of smoldering (indolent) rather than an aggressive form of MM.2 Although derived from peripheral blood, ANBL-6 recapitulates many of the phenotypes associated with primary plasma cell tumors in the bone marrow. ANBL-6 is a highly representative cellular model of multiple myeloma.
Application
- Each vial contains >1X106 viable cells.
- Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
- Cells are verified to be of human origin and negative for inter-species contamination from mouse, rat, chinese hamster, Golden Syrian hamster, and Non-human Primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
- Cells are negative for mycoplasma contamination.
Multiple myeloma (MM) is a cancer of the plasma cells originating in bone marrow. Although a rare cancer, there is no cure and only 54% of patients diagnosed with MM survive beyond 5 years.1 The vast majority of MM cases are known as smoldering or asymptomatic MM, wherein a small population of aberrant plasma cells resides in the bone marrow.2 Most MM cell line models are not derived from originating tissue but rather peripheral blood, raising questions of how closely these lines reflect the characteristics of the disease in the originating hematopoietic environment.2 SourceThe ANBL-6 cell line was isolated from peripheral blood of a 67-year old female patient with myeloma.3References1. Cancer Statistics: Multiple Myeloma https://www.cancer.net/cancer-types/multiple-myeloma/statistics Science 2004; 306(5694): 269-71.2. Leukemia 2020; 34(10): 2754-2765.3. Cancer Res 1993; 53(21): 5320-5327.
Features and Benefits
ANBL-6 is a highly representative cellular model of multiple myeloma.
Storage and Stability
ANBL-6 Human Plasma Cell Myeloma Cell Line should be stored in liquid nitrogen until use. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.
Other Notes
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service