Skip to Content
Merck
All Photos(4)

Key Documents

SCC165

Sigma-Aldrich

BLaER1 Human B-cell Precursor Leukemia Cell Line

Human

Synonym(s):

B cell Leukemia C/EBPalphaER clone 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

Product Name

BLaER1 Human B-cell Precursor Leukemia Cell Line, BLaER1 human B-cell precursor leukemia cell line may be transdifferentiated and used to model human monocytes.

biological source

human

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

General description

Induction of differentiation is a therapeutic approach for cancer. Treatment of leukemias with differentiation-inducing factors in combination with pro-apoptotic compounds has been shown to be highly effective . Ectopic expression of the transcription factor C/EBPa causes reprogramming of leukemia cells, resulting in expression of a normal cell phenotype.

The BLaER1 cell line is a tamoxifen-inducible derivative of the RCH-ACV B-cell leukemia cell line genetically modified using a retrovirus vector to express C/EBPa fused with the estrogen receptor hormone binding domain (ER) and GFP . A study demonstrated that BLaER1 cells could efficiently convert into cells exhibiting increased adherent, phagocytic and quiescent properties with a transcriptome resembling normal macrophages . Cells retained their phenotype even when C/EBPa was inactivated, a characteristic of genuine cell reprogramming. C/EBPa induction also impaired cell tumorigenicity after transplantation into immunodeficient mice. BLaER1 is thus a model for highly efficient transdifferentiation of B-cell-derived human cancer cells.

Source:
BLaER1 cells are a single subclone isolated from C/EBPaER-GFP-transfected RCH-ACV acute lymphoblastic leukemia (ALL) cells sorted for GFP expression. The parental cell line was obtained from the bone marrow of a female with chromosome translocation 1;19 and trisomy 8 .

Biosafety Level 2:
BLaER1 cells contain squirrel monkey retrovirus (SMRV) and should be handled under biosafety level 2.

Cell Line Description

Cancer Cells

Application

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Quality

  • Each vial contains ≥1×10 viable cells.
  • Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells are verified to be of human origin and negative for inter-species contamination from mouse, rat, chinese hamster, Golden Syrian hamster, and non-human primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
  • Cells are negative for mycoplasma contamination.
  • Cells tested positive for squirrel monkey retrovirus (SMRV).

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Waxman
Leukemia, 14(3), 491-496 (2000-03-17)
Successful treatment of acute promyelocytic leukemia (APL) has identified several novel approaches to induce leukemic cell differentiation and selective apoptosis by overcoming the site-specific transcriptional repression by dominant fusion leukemogenic proteins characteristic of APL and other forms of acute myelogenous
Bowen Zhou et al.
Science signaling, 15(765), eabl6781-eabl6781 (2022-12-21)
Pyroptosis is a mechanism of programmed, necrotic cell death mediated by gasdermins, a family of pore-forming proteins. Caspase-1 activates gasdermin D (GSDMD) under inflammatory conditions, whereas caspase-3 activates GSDME under apoptotic conditions, such as those induced by chemotherapy. These pathways
C/EBPα Induces Highly Efficient Macrophage Transdifferentiation of B Lymphoma and Leukemia Cell Lines and Impairs Their Tumorigenicity.
Francesca Rapino et al.
Cell reports, 19(6), 1281-1281 (2017-05-13)
I Jack et al.
Cancer genetics and cytogenetics, 19(3-4), 261-269 (1986-01-15)
A cell line (RCH-ACV) was established from a bone marrow sample of a child with acute lymphoblastic leukemia (ALL). The cell line lacked Epstein-Barr virus nuclear antigen and exhibited a recently described nonrandom chromosome translocation, 1;19, thought to be associated
Francesca Rapino et al.
Cell reports, 3(4), 1153-1163 (2013-04-03)
Earlier work demonstrated that the transcription factor C/EBPα can convert immature and mature murine B lineage cells into functional macrophages. Testing >20 human lymphoma and leukemia B cell lines, we found that most can be transdifferentiated at least partially into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service