Skip to Content

Fechas de fin de año: Facturación de pedidos hasta 13/12. Receso de Merck Argentina del 23/12 al 02/01.

Merck
All Photos(1)

Key Documents

S7860

Sigma-Aldrich

CpGenome Rat Unmethylated Genomic DNA Standard

Useful as a control or standard for variety of epigenomic assays such as methylated or unmethylated DNA enrichment, bisulfite conversion or melt curve analysis to determine the status of global or locus- specific DNA methylation.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

Pricing and availability is not currently available. Contact Technical Service

Quality Level

species reactivity

rat

manufacturer/tradename

CpGenome
Upstate®

shipped in

wet ice

Related Categories

General description

The CpGenome Rat Unmethylated Genomic DNA Standard is ideal for use as a control or standard in DNA methylation studies. This unmethylated standard is generated using multiple strand displacement amplification of rat genomic DNA using Phi29 DNA polymerase. This process results in a DNA standard which contains ≥ 95% unmethylated cytosines across the genome in both heterochromatic and euchromatic regions.

Application

Research Category
Epigenetics & Nuclear Function

Quality

Quantitative Methylation Specific PCR (qMSP):
Bisulfite converted unmethylated and methylated rat genomic DNA are assessed for methylation status at various loci within the genome.

Physical form

5 µg DNA in buffer containing 10mM Tris-HCl pH 8.0, 0.1mM EDTA

Storage and Stability

Upon receipt aliqout and store at -20°C to avoid multiple freeze thaws which can fragment DNA. For best quality, limit freeze and thaw of the same –20°C DNA aliquot to no more than 3 times. For longer term storage, the DNA can be stored at -80°C. With proper storage and protection from excess freeze-thaw cycles, this product is stable for up to two years from the date of receipt.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service