Skip to Content
Merck
All Photos(1)

Key Documents

RDYSTRAIN01

Millipore

ReadyStream® Mesh Strainer

Mesh Strainer to be used as needed to reduce particles upstream of ReadyStream®

Synonym(s):

Particle Strainer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13102014
NACRES:
NB.51

Pricing and availability is not currently available.

packaging

pkg of 1 unit

system L × W × H

21 cm × 10 cm × 5 cm

application(s)

food and beverages

compatibility

for use with ReadyStream® System

storage temp.

15-25°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
5.74826RDYSTRM50RDYSTRM60
application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

packaging

pkg of 1 unit

packaging

pack of 5 filter

packaging

pkg of 1 unit

packaging

pkg of 1 unit

storage temp.

15-25°C

storage temp.

-

storage temp.

room temp

storage temp.

room temp

system L × W × H

21 cm × 10 cm × 5 cm

system L × W × H

-

system L × W × H

-

system L × W × H

-

General description

ReadyStream® Mesh Strainer is an accessory to be used with the ReadyStream® System. The mesh strainer ensures reduction of particles directed towards the ReadyStream® instrument enhancing the performance and life span of the equipment.

Application

ReadyStream® Mesh Strainer is designed to act as a pre-filter fitter upstream to the ReadyStream® instrument.

Legal Information

READYSTREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service