Skip to Content
Merck
All Photos(1)

Key Documents

PRCYTA-40K

Millipore

MILLIPLEX® Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A

Synonym(s):

Luminex® non-human primate cytokine immunoassay, Millipore non-human primate cytokine panel, Monkey cytokine multiplex kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.47


To order a Milliplex® kit, please search for your analyte of interest.

species reactivity

nonhuman primates

technique(s)

multiplexing: suitable

assay range

inter-assay cv: <20%
intra-assay cv: <10%

inter-assay cv: <30%
(TFN-α)

detection method

fluorometric (Luminex® xMAP®)

shipped in

wet ice

storage temp.

2-8°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
BP1073PHR1887Y0000238
manufacturer/tradename

USP

manufacturer/tradename

BP

manufacturer/tradename

-

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

-

format

neat

API family

perindopril

API family

perindopril

API family

perindopril

API family

perindopril

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

General description

Cytokine is a general term used for a diverse group of small proteins, peptides or glycoproteins secreted by lymphocytes, monocytes, macrophages and other cells that regulate immune responses, haematopoiesis and lymphocyte development. Cytokines include interleukins, interferons, chemokines and other signaling molecules. Growth factors are extracellular polypeptides that have a positive effect on cell growth and proliferation. They can stimulate growth in a variety of different cell types. Growth factors and cytokines are similar in structure and the way of action. Individual cytokines or growth factors are produced by multiple cell types. This is different from hormones, which tend to be made by specialized glands. Each cytokine or growth factor acts through its own receptor on target cells to generate signaling pathways, and as a consequence to regulate biological processes. Some intracellular signaling components are shared between cytokines and growth factors. Expression of cytokines or growth factors and their receptors is highly regulated. De-regulation may contribute to many diseases such as infectious disease, autoimmune and chronic inflammatory disease, cardiovascular disease, metabolic syndrome, neurological disorders and cancer. Cytokine and growth factor research plays a significant role in achieving a deeper understanding of the immune system and combating related diseases. As the most closely related species to humans, non-human primates are critical models for those studies.
The MILLIPLEX® Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A 48-Plex Premixed Magnetic Bead Panel is a 48-plex kit to be used for the simultaneous quantification of the following analytes in serum or plasma samples and tissue/cell lysate and culture supernatant samples: BCA-1, sCD137, sCD40L, Eotaxin, sFASL, FGF-2, Fractalkine, G-CSF, GM-CSF, Granzyme A, Granzyme B, IFNα2, IFNγ, IL-1α,
IL-1β, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p70), IL-15, IL-16, IL-17A, IL-17E, IL-18, IL-21, IL-22, IL-23, IL-28A, IL-31, IL-33, IP-10, I-TAC, MCP-1, MIG, MIP-1α, MIP-1β, MIP-3α, Perforin, RANTES, TGFα, TNFα, TNFβ, VEGF-A.

Specificity

There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes in this panel, except G-CSF had ∼5% cross-reactivity with IL-1β.

Application

  • Analytes: BCA-1, sCD137, CD40L, Eotaxin, sFASL, FGF-2, Fractalkine, G-CSF, GM-CSF, Granzyme A, Granzyme B, IFNα2, IFNγ, IL-1α, IL-1β, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p70), IL-15, IL-16, IL-17A, IL-17E, IL-18, IL-21, IL-22, IL-23, IL-28A, IL-31, IL-33, IP-10, I-TAC, MCP-1, MIG, MIP-1α, MIP-1β, MIP-3α, Perforin, RANTES, TGFα, TNFα, TNFβ, VEGF-A
  • Recommended Sample type: Serum, plasma or tissue/cell lysate and culture supernatant samples
  • Recommended Sample dilution: Neat plasma or serum. Tissue culture supernatant may require a dilution with an appropriate control medium prior to assay.
  • RANTES requires a 1:100 dilution of plasma/serum samples.
  • NOTE: RANTES cannot be plexed with other cytokines in this panel due to a required 1:100 dilution of plasma/serum samples.
  • Note: The serum/plasma samples of some non-human primate species may require up to 1:100 dilution in Assay Buffer for IL-8.
  • Assay Run Time: Overnight or two-hour primary incubation. For best results, an overnight incubation is recommended.
  • Research Category: Inflammation & Immunology
  • Research Sub Category: Obesity, Metabolic Disorders, Inflammation & Autoimmune Mechanisms.

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Storage and Stability

  • Recommended storage for kit components is 2−8°C.
  • For long-term storage, freeze reconstituted standards and controls at −20°C. Avoid multiple freeze/thaw cycles.
  • DO NOT FREEZE Antibody-Immobilized Beads, Detection Antibody, and Streptavidin-Phycoerythrin.

Analysis Note

  • This is an overnight or two-hour incubation assay.
  • This assay requires 25 μL of neat plasma or serum (1:100 dilution for RANTES) or 25 μL cell culture supernatant per well.
  • This kit may be used for the analysis of all of the Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A analytes in human tissue/cell lysate and culture supernatant samples and serum or plasma samples.
  • RANTES cannot be plexed with other cytokines in this panel due to a required 1:100 dilution of plasma/serum samples. For more information, please carefully review the sample preparation for serum and plasma samples and the preparation of antibody-immobilized beads sections below when assaying RANTES.
  • The serum/plasma samples of some non-human primate species may require up to 1:100 dilution in Assay Buffer for IL-8.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Related Content

    MILLIPLEX® Non-Human Primate Panel A allows comprehensive immune factor profiling on a single 96-well plate.

    MILLIPLEX® Non-Human Primate Panel A allows comprehensive immune factor profiling on a single 96-well plate.

    MILLIPLEX® Non-Human Primate Panel A allows comprehensive immune factor profiling on a single 96-well plate.

    MILLIPLEX® Non-Human Primate Panel A allows comprehensive immune factor profiling on a single 96-well plate.

    See All

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service