Skip to Content
Merck
All Photos(1)

Key Documents

OP15

Sigma-Aldrich

Anti-c-ErbB2/c-Neu (Ab-3) Mouse mAb (3B5)

liquid, clone 3B5, Calbiochem®

Synonym(s):

Anti-HER2, Anti-c-Neu, Anti-ErbB2, Anti-Erythroblastosis Virus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3B5, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, mouse

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ERBB2(2064)

General description

Anti-c-ErbB2/c-Neu (Ab-3), mouse monoclonal, clone 3B5, recognizes the ~185 kDa c-ErbB2/c-Neu protein in HEK293 cells. It is validated for WB, IF, IP, and IHC on frozen and paraffin sections.
Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with Sp2/0.Ag14 cells (see application references). Recognizes the ~185 kDa c-Erb2/c-Neu protein.
Recognizes the ~185 kDa c-ErbB2/c-Neu protein in HEK293 cells.

Immunogen

Human
a synthetic peptide (TAENPEYLGLDVPV) corresponding to amino acids 1242-1255 from the C-terminal domain of human c-ErbB2/c-Neu

Application


Frozen Sections (2.5 g/ml)
Immunoblotting (2-3 g/ml, see application references)
Immunofluorescence (2.5 g/ml)
Immunoprecipitation (1 g/sample, see application references)
Paraffin Sections (2.5 g/ml, pressure cooker, heat or trypsin pre-treatment required, see application references)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 0.05 M sodium phosphate buffer, 0.2% gelatin.

Analysis Note

Negative Control
HepG2 cells or normal skin
Positive Control
SK-BR-3 cells or breast carcinoma tissue

Other Notes

For paraffin sections, we recommend pre-treating with a pressure cooker, but trypsin and heat will also work well. Including 0.05% saponin during staining of paraffin sections may help reduce background. In one study ductal carcinomas in situ displaying large-cell, comedo growth type, were stained while none of 16 ductal carcinomas in situ of small-cell, papillary, or cribriform growth type were stained by this antibody. Antibody should be titrated for optimal results in individual systems.
Mamcs, J.R., et al. 1994. Ann. Surg.219, 332.
Tetu, B., et al. 1994. Cancer73, 2359.
DiFiore, P.P., et al. 1987. Science237, 178.
Slamon, D.J., et al. 1987. Science235, 177.
Varley, J.M., et al. 1987. Oncogene1, 423.
Bargmann, C.I., et al. 1986. Nature319, 226.
Yamamoto, T., et al. 1986. Nature319, 230.
Blick, M., et al. 1984. Blood64, 1234.
Schwab, M., et al. 1984. Cold Spring Harbor Laboratory 2, 215.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Henriette O'Geen et al.
Epigenetics & chromatin, 12(1), 26-26 (2019-05-06)
Rewriting of the epigenome has risen as a promising alternative to gene editing for precision medicine. In nature, epigenetic silencing can result in complete attenuation of target gene expression over multiple mitotic divisions. However, persistent repression has been difficult to
Peter Nagy et al.
Journal of cell science, 115(Pt 22), 4251-4262 (2002-10-12)
The ErbB family of transmembrane receptor tyrosine kinases plays an important role in the pathogenesis of many cancers. The four members of the family, ErbB1-4, form various homo- and heterodimers during the course of signal transduction. A second hierarchical level
Zongming Liu et al.
Scientific reports, 9(1), 622-622 (2019-01-27)
Breast cancer brain metastasis (BCBM) remains a major clinical problem. Approximately 10-16% of patients with breast cancer develop brain metastases (BCBM). However, no systemic therapy has gained regulatory approval for the specific treatment of BCBM and this remains an area
Yoriko Yamashita-Kashima et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 17(15), 5060-5070 (2011-06-28)
We investigated the antitumor activity of the combination of two different humanized monoclonal human epidermal growth factor receptor (HER) 2 antibodies, pertuzumab and trastuzumab, for gastric cancer. Tumor mouse xenograft models were used to examine antitumor activity. Cell proliferation was
Robin Kuns et al.
Oncogene, 24(43), 6502-6515 (2005-07-12)
4.1B is a member of the protein 4.1 superfamily of proteins that link transmembrane proteins to the actin cytoskeleton. The 4.1B gene localizes to chromosome 18p11.3, which undergoes loss of heterozygosity in mammary tumors. Here, we examine the expression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service