Skip to Content
Merck
All Photos(1)

Key Documents

MXSMTLP48

Millipore

Milliflex® Pre-filled Media Cassettes

Tryptic Soy Agar, Polysorbate 80, Lecithin, for heterotrophic bacteria, pack of 48, suitable for bioburden testing

Synonym(s):

Tryptic Soy Agar with Polysorbate 80 and Lecithin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
eCl@ss:
32160801
NACRES:
NA.74

Agency

International standards

Quality Level

sterility

aseptically filled

form

plate (pre-poured agar)

shelf life

12 mo.

feature

ready-to-use

composition

Lecithin
Polysorbate 80

packaging

cassette of 10 mL
pack of 48

manufacturer/tradename

Milliflex Oasis®

color

clear tan to light amber

input

sample type pharmaceutical(s)
sample type cosmetics
sample type beer
sample type bottled water

pH

7.3±0.2 (25.0 °C)

suitability

suitable for (suitable for solid cultivation)
heterotrophic bacteria

application(s)

bioburden testing
cosmetics
microbiology
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® System

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Milliflex® Cassette are ready to use agar-based media to be used with Milliflex® or Milliflex Oasis® system for convenient, rapid and reliable bioburden testing. The prefilled agar cassette helps to streamline the microbial contamination testing workflow with quality assurance and avoiding risks of exogenous contamination. Each plate is a 3-piece assembly containing approximately 10ml of the media. Tryptic Soy Agar is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The medium is supplemented with the neutralizers such as lecithin and polysorbate 80 to neutralize for quaternary ammonium compounds and substituted phenolic disinfectants respectively.

Application

Milliflex® Cassette prefilled with Tryptic Soy Agar is used in Bioburden testing and general microbiological analysis. It is commonly used in determining the efficiency of the sanitation of containers, equipment and surfaces as well as for water miscible cosmetic products. It is widely used in pharmaceutical, cosmetic and brewing industry.

Culture Medium

Trypticase Soy w/Poly 80 and Lecithin

Analysis Note

Colonies appear clear to creamy white, some may produce pigment
QC organism
E. coli (ATCC 25922)
S. aureus (ATCC 6538)
Bacillus spizizenii ATCC 6633
Pseudomonas aeruginosa ATCC 9027
Candida albicans ATCC 10231
Aspergillus brasiliensis ATCC 16404
Target Microorganisms: Heterotrophic microorganisms

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service