Skip to Content
Merck
All Photos(2)

Key Documents

MT17MAG47K-PX25

Millipore

MILLIPLEX® Mouse TH17 Magnetic Bead Panel - Premixed 25 plex - Immunology Multiplex Assay

Simultaneously analyze multiple Th17 cytokine and chemokine biomarkers with the Th17 Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.

Synonym(s):

Mouse TH17 Panel, TH17 Magnetic Bead Panel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

Quality Level

species reactivity

mouse

manufacturer/tradename

Milliplex®

assay range

accuracy: 85-110%
linearity: 111.0%
(1:04)

linearity: 113.1%
(1:08)

standard curve range: 1.5-1,500 pg/mL
(IL-4)

standard curve range: 10-10,000 pg/mL
(IL-17F)

standard curve range: 127-130,000 pg/mL
(IL-28B)

standard curve range: 15-15,000 pg/mL
(IL-1β)

standard curve range: 2.4-2,500 pg/mL
(IL-22)

standard curve range: 20-20,000 pg/mL
(IL-10, IL-12(p70), & IL-21)

standard curve range: 3.4-3,500 pg/mL
(TNFα)

standard curve range: 34-35,000 pg/mL
(GM-CSF & IL-15)

standard curve range: 342-350,000 pg/mL
(IL-23)

standard curve range: 39-40,000 pg/mL
(IL-13 & IL-17A)

standard curve range: 4.9-5,000 pg/mL
(IL-5)

standard curve range: 488-500,000 pg/mL
(TNFβ)

standard curve range: 49-50,000 pg/mL
(CD40L IL-31 & MIP-3α)

standard curve range: 586-600,000 pg/mL
(IL-17E/ IL-25)

standard curve range: 6.9-6,000 pg/mL
(IL-2)

standard curve range: 7.8-8,000 pg/mL
(IFNγ & IL-6)

standard curve range: 78-80,000 pg/mL
(IL-33)

standard curve range: 879-900,000 pg/mL
(IL-27)

technique(s)

multiplexing: suitable

compatibility

configured for Premixed

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

CD4 T-helper cells are major players in adaptive immunity. Based on their expression profile of transcription factors and secreted cytokines, these cells are divided into the major subsets Th1, Th2, Th17 and T regulatory (Treg) cells. Th1 cells, induced by IL-12 and enhanced by IFNγ, secrete IFNγ that activates macrophages. Induced by IL-4, Th2 cells primarily produce IL-4, IL-5, IL-13, IL-25 and IL-17E, playing a role in allergy. Treg cells, induced by TGFβ, produce the anti-inflammatory cytokines IL-10 and TGFβ; and function to control T-cell responses to prevent autoimmune reactions. Th17 cells, a more recently discovered subset characterized by the production of IL-17, are induced by TGFβ; combined with IL-23 and IL-1β; in humans, and TGFβ, IL-6 and IL-21 in mice. Activated Th17 cells secrete IL-17A, IL-17F, IL-21, IL-22, and TNFα; to promote tissue inflammation.

Th17 cells are involved in the clearance of extracellular bacteria and fungi. They are abundant in the intestinal lamina propria and function as a barrier against invading pathogens. Excessive amounts of Th17 cells have been implicated in the pathogenesis of several autoimmune diseases, including multiple sclerosis, psoriasis, juvenile diabetes, rheumatoid arthritis, Crohn′s disease, and autoimmune uveitis. In addition, Th17 cells play an important role in tumor development, progression, and metastasis, potentially in both promoting and inhibiting tumor growth.

The MILLIPLEX® Mouse Th17 Bead panel enables you to focus on the therapeutic potential of the modulation of CD4 T-helper cell cytokines. Coupled with the Luminex® xMAP®platform in a bead format, you receive the advantage of ideal speed and sensitivity, allowing quantitative multiplex detection of dozens of analytes simultaneously, which can dramatically improve productivity.

Panel Type: Cytokines/Chemokines

Specificity

Cross Reactivty
There was no or minimum cross-reactivity between the antibodies and any other analytes in the panel (see details in assay protocol).
Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • Analytes: CD40 Ligand, GM-CSF, IFN-γ, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-10, IL-12 (p70), IL-13, IL-15, IL-17A, IL-17E/IL-25, IL-17F, IL-21, IL-22, IL-23, IL-27, IL-28B, IL-31, IL-33, MIP-3α/CCL20, TNF-α, TNFβ
  • Recommended Sample type: serum, plasma or tissue/cell lysate and culture supernatant
  • Recommended Sample dilution: neat
  • Assay Run Time: overnight or one day
  • Research Category: Inflammation & Immunology

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: Refer to kit protocol for sensitivities of individual biomarkers.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sansana Sawasdikosol et al.
ImmunoHorizons, 4(7), 382-391 (2020-07-08)
Hematopoietic progenitor kinase 1 (HPK1) is a negative regulator of TCR-initiated signal transduction. Both the HPK1-/- mice and the genetically engineered mice with a point mutation that disrupts the catalytic activity of HPK1 possess enhanced antitumor immunity, especially when these
Mab Pereira Corrêa et al.
Journal of molecular medicine (Berlin, Germany), 95(9), 1005-1015 (2017-07-01)
Atopic dermatitis (AD) is caused by both dysregulated immune responses and an impaired skin barrier. Although beta-galactoside-binding protein galectin-1 (Gal-1) has immunomodulatory effects in several inflammatory disorders, therapeutic strategies based on its anti-inflammatory properties have not been explored in AD.
Frans Eberth Costa Andrade et al.
The British journal of ophthalmology, 102(7), 1003-1010 (2018-03-05)
To evaluate galectin-3 (Gal-3), a β-galactoside binding protein, as a possible biomarker in ocular allergy and further investigated the role of endogenous Gal-3 in a murine model of ovalbumin (OVA)-induced allergic conjunctivitis (AC). Conjunctival impression cytology specimens from control and
Li-Rong Li et al.
Molecular medicine (Cambridge, Mass.), 22, 800-808 (2016-11-24)
Glucagon-like peptide 1 (GLP-1) can promote islet β-cell replication and function, and mesenchymal stem cells (MSCs) can inhibit T cell autoimmunity. This study aimed at testing the dynamic distribution of infused human MSCs and therapeutic effect of combined MSCs and
Lirong Li et al.
Scientific reports, 6, 37894-37894 (2016-12-03)
Mesenchymal stem cells (MSCs) transplantation is a promising therapeutic strategy for type 1 diabetes (T1D). However, little is known on whether MSC transplantation can benefit T1D patients with ketoacidosis and its potential actions. Here, we show that infusion with bone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service