MMPWRSPIN
Milliflex Oasis® power supply
India adaptor – to power supply up to three Milliflex Oasis® filtration pumps
Synonym(s):
Power Supply
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116129
NACRES:
NB.79
Recommended Products
AC/DC input
100 V / 250 V, 50-60 Hz
packaging
pkg of 1 ea
manufacturer/tradename
Milliflex Oasis®
application(s)
bioburden testing
food and beverages
pharmaceutical
water monitoring
compatibility
India adaptor – to power supply up to three Milliflex Oasis® filtration pumps
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
MILLIFLEX OASIS® filtration system offers a solution to simplify and secure microbial testing process with premium levels of productivity, result reliability and regulatory compliance. The MILLIFLEX OASIS® pump is a new generation vacuum pump for water and bioburden analysis in pharmaceutical environments. A power supply of Indian standard with 100 - 240 V, 50/60 enables efficient functioning of the filtration pump. One set of MILLIFLEX OASIS® power supply can be used to power up to three pumps.
Application
MILLIFLEX OASIS® filtration pump finds its application in bioburden testing and water monitoring of beer, bottled water, cosmetics and Pharmaceutical Products Analysis.
Legal Information
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service