Skip to Content
Merck
All Photos(2)

Key Documents

MMPWRSPEU

Millipore

Milliflex Oasis® power supply

Europe adaptor – to power supply up to three Milliflex Oasis® filtration pumps

Synonym(s):

Power supply

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.79
Pricing and availability is not currently available.

AC/DC input

100 V / 250 V, 50-60 Hz

Quality Level

packaging

pkg of 1 ea

manufacturer/tradename

Milliflex Oasis®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

Europe adaptor – to power supply up to three Milliflex Oasis® filtration pumps

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

MILLIFLEX OASIS® filtration system offers a solution to simplify and secure microbial testing process with premium levels of productivity, result reliability and regulatory compliance. MILLIFLEX OASIS® pump is a new generation vacuum pump for water and bioburden analysis in pharmaceutical environments. A power supply of European standard with 100 - 240 V, 50/60 enables efficient functioning of the filtration pump. One set of MILLIFLEX OASIS® power supply can be used to power up to three pumps.

Application

MILLIFLEX OASIS® filtration pump finds its application in bioburden testing and water monitoring of beer, bottled water, cosmetics and Pharmaceutical Products Analysis.

Legal Information

MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service