Skip to Content
Merck
All Photos(1)

Key Documents

MMA130

Millipore

Millicell® µ-Angiogenesis Activation Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.76

species reactivity

rat, vertebrates, human

technique(s)

activity assay: suitable
cell culture | mammalian: suitable

detection method

fluorometric

General description

Angiogenesis, the formation of new blood vessels from a pre-existing vascular network, occurs normally in development and is critical for a majority of the vessel formation that occurs during embryogenesis, tissue generation, and wound healing. However, abnormal blood vessel growth, either excessive or insufficient, can be the underlying cause of many deadly and debilitating diseases including cancer, cardiovascular disease, stroke, and diabetic and age-related blindness. Identification of specific compounds that promote or inhibit angiogenesis may provide promising treatments for these diseases.

Tubular formation is a multi-step process involving cell adhesion, migration, differentiation, and growth. Millipore′s TM=["Millicell"] µ-Angiogenesis Inhibition Assay provides an efficient system for the rapid and accurate identification of factors that inhibit tube formation of endothelial cells. The µ-Angiogenesis slide, with enhanced optical capabilities in a microscale format, allows for real time and continuous monitoring of tube formation in a simplifed and reproducible manner.

Components

1. Millicell® µ-Angiogenesis Slides: (Part No. CS203030) Five tissue culture treated slides. Each slide contains 15 wells. 2. Fibrinogen Solution: (Part No. CS203035) One 1.5 mL vial. 3. Thrombin Solution: (Part No. CS203036) One 0.75 mL vial. 4. ITS Media Supplement, 100X: (Part No. CS203037) One 0.5 mL vial. 5. PMA (Phorbol 12-Myristate 13-Acetate): (Part No. CS203029): One 1 mg vial. 6. Calcein-AM: (Part No. CS202541) One 50 µg vial.

Storage and Stability

1. Millicell® µ-Angiogenesis Slides (Part No. CS203030) should be stored at room temperature until ready to use.2. Fibrinogen Solution (Part No. CS203035) should be stored at -20°C and used within six months from the date of receipt. For short-term storage, store at room temperature for up to 2 weeks. Do not store at 4°C, as Fibrinogen Solutions tend to form aggregates when stored at 4°C. For long-term storage, aliquot into sterile tubes in a sterile laminar flow hood, freeze on dry ice, and store at -20°C.3. Thrombin Solution (Part No. CS203036) should be stored at -20°C and used within six months from the date of receipt. Unused Thrombin Solution can be stored at 4°C for up to 1 month. For longer term storage, Thrombin Solution may be stored in aliquots at -20°C.4. ITS Media Supplement, 100X (Part No. CS203037) should be stored at -20°C. Unused portions may also be stored at 4°C for up to 1 year. Avoid freeze thaw cycles.5. PMA (Phorbol-12-Myristate-13-Acetate) (Part No. CS203029) is provided as a 1 mg lyophilized powder. To prepare a 1 mg/mL stock, add 1 mL sterile DMSO (not provided). Vortex vigorously. Centrifuge briefly to collect the solution. Unused PMA Solution may be stored at -20°C in the dark for at least six months. WARNING: Highly irritating to skin and mucous membranes. TUMOR PROMOTER. Read the Material Safety Data Sheet (MSDS) prior to rehydration and use. Gloves and mask should be worn when using this compound.6. Calcein AM (Part No. CS202541) is provided as a 50 µg lyophilized powder. To prepare 1 mM Calcein AM Stock Solution, briefly centrifuge before opening the tube and then add 40 µL sterile DMSO (not provided). Vortex vigorously. Centrifuge briefly to collect the solution. Unused Calcein AM Solution may be stored in aliquots at –20°C for up to 1 month.

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service