Skip to Content
Merck
All Photos(1)

Key Documents

MABT844

Sigma-Aldrich

Anti-Cytokeratin 8/18 Antibody, clone L2A1

clone L2A1, from mouse

Synonym(s):

Cytokeratin-8/18, Keratin type II cytoskeletal 8/Keratin type I cytoskeletal 18, Keratin-8/Keratin-18, K8/K18)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

Pricing and availability is not currently available.

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

L2A1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgGκ

UniProt accession no.

target post-translational modification

unmodified

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
B7653SAB4200680A0185
biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

antibody form

purified immunoglobulin

antibody form

ascites fluid

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

conjugate

agarose conjugate

conjugate

unconjugated

conjugate

-

conjugate

peroxidase conjugate

shipped in

wet ice

shipped in

dry ice

shipped in

dry ice

shipped in

-

technique(s)

ELISA: suitable, immunoprecipitation (IP): suitable, immunohistochemistry: suitable, western blot: suitable

technique(s)

capture ELISA: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, indirect ELISA: 1:4,000

technique(s)

ELISA: suitable, immunocytochemistry: suitable, immunoprecipitation (IP): suitable, indirect ELISA: 0.15-0.3 μg/mL, western blot: suitable

technique(s)

dot blot: suitable, flow cytometry: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:300, indirect ELISA: 1:30,000, western blot (chemiluminescent): 1:80,000

General description

Cytokeratin-8/18 (UniProt: P05787/P05783; also known as Keratin, type II cytoskeletal 8/ Keratin, type I cytoskeletal 18, Keratin-8/Keratin-18, K8/K18) is encoded by the KRT8/KRT18 (also known as CYK8/CYK18) gene (Gene ID: 3856/3875) in human. Cytokeratins 8 and 18 (K8/18) are simple epithelial cell-specific intermediate filament proteins. Cytokeratin-8 is a type II, neutral to basic, protein of the intermediate filament family that together with Cytokeratin-19 (KRT19) helps to link the contractile apparatus to dystrophin at the costameres of striated muscle. Cytokeratin-8 can undergo phosphorylation on three major serine residues: Serine 23, 431, and 73. Serine 23 is shown to be highly conserved in all type II keratins. Phosphorylation at Serine 73 is reported to increase during cellular stress, including hear and drug exposure. However, under normal conditions serine 73 remains largely dephosphorylated. Cytokeratin-8 can also undergo O-glycosylation in a cell cycle-dependent manner and glycosylation increases its solubility and reduces stability by inducing proteasomal degradation. Cytokeratin-18 is a type I, acidic, protein that is expressed in colon, placenta, and liver. Higher expression levels have been reported in lymph nodes of breast carcinoma. Cytokeratin-18 is involved in the uptake of thrombin-antithrombin complexes by hepatic cells. Upon phosphorylation, it plays a role in filament reorganization. It is proteolytically cleaved by caspases during epithelial cell apoptosis and this cleavage is shown to occur at Asp-238. Phosphorylation of cytokeratin-18 at serine 33 is shown to increase during mitosis in cultured cells and in regenerating liver. Serine 33 phosphorylation is considered to be essential for its association with 14-3-3 proteins and has a role in keratin organization and distribution. Cytokeratin-18 can undergo O-glycosylation, which results in its increased solubility and reduction in its stability. Mutations in KRT8 and KRT18 genes have been been linked to liver cirrhosis that is characterized by severe panlobular liver-cell swelling with Mallory body formation, prominent pericellular fibrosis, and marked deposits of copper.

Specificity

Clone L2A1 is a mouse monoclonal antibody that detects human cytokeratin 8/18.

Immunogen

KLH-conjugated Linear Peptide corresponding to a sequence from Human Cytokeratin 8/18.

Application

Anti-Cytokeratin 8/18, clone L2A1, Cat. No. MABT844, is a mouse monoclonal antibody that detects cytokeratin 8/18 and has been tested for use in Immunocytochemistry and Immunoprecipitation.
Immunoprecipitation Analysis: A representative lot detected Cytokeratin 8/18 in Immunoprecipitation applications (Haltiwanger, R.S., et. al. (1997). J Biol Chem. 272(13):8752-8; Chou, C.F., et. al. (1994). Biochem J. 298 ( Pt 2):457-63; Chou, C.F., et. al. (1993). Biochem J. 268(6):4465-72; Ku, N.O., et. al. (2000). J Cell Biol. 149(3):547-52; Lahdeniemi, I.A.K., et. al. (2017). Cell Death Differ. 24(6):984-996).

Immunocytochemistry Analysis: A representative lot detected Cytokeratin 8/18 in Immunocytochemistry applications (Chou, C.F., et. al. (1993). Biochem J. 268(6):4465-72).
Research Category
Cell Structure

Quality

Evaluated by Immunocytochemistry in HT-29 cells.

Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected Cytokeratin 8/18 in HT-29 cells.

Target description

~53.70 kDa and 48.06 kDa, respectively for cytokeratin 8 and 18 calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service