Skip to Content
Merck
All Photos(2)

Key Documents

MABT51

Sigma-Aldrich

Anti-Galectin-3 Antibody, clone M3/38

clone M3/38, from rat

Synonym(s):

lectin, galactoside-binding, soluble, 3, Mac-2 antigen, Carbohydrate-binding protein 35, galectin-3, Galactose-specific lectin 3, Laminin-binding protein, Lectin L-29, IgE-binding protein, Galactoside-binding protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

M3/38, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LGALS3(3958)

General description

Galectin-3 belongs to the galectin gene family defined by the specificity of a carbohydrate recognition domain (CRD) for Gal/Lac and expression is usually elevated in a wide range of neoplastic cell types. The galectin CRD consists of 135 amino acids. Galectin-3 has been shown to effect tumor development and progression.

Immunogen

Plasma membrane glycoproteins corresponding to mouse Galectin-3.

Application

Anti-Galectin-3 Antibody, clone M3/38 is an antibody against Galectin-3 for use in WB, IP, IH & IC.
Western Blot Analysis: 1:50,000 dilution from a previous lot detected Galectin-3 in 10 µg of NIH/3T3 cell lysate.

Immunoprecipitation Analysis: A previous lot was used by an independent laboratory in IP. (Springer, T., et al. (1981). THE JOURNAL OF BIOLOGICACHLE MISTRY. 256(8):3833-3839.)

Immunohistochemistry Analysis: A previous lot was used by an independent laboratory in IH. (Gasbarri, A., et al. (1999). Journal of Clinical Oncology. 17(11):3494-3502.)

Immunocytochemistry Analysis: A previous lot was used by an independent laboratory in IC. (Gasbarri, A., et al. (1999). Journal of Clinical Oncology. 17(11):3494-3502.)

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 1:50,000 dilution of this antibody detected Galectin-3 in 10 µg of HeLa cell lysate.

Target description

~28 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie-Jen Lee et al.
International journal of endocrinology, 2021, 5583491-5583491 (2021-05-27)
Accumulating evidence suggests that galectin-3 is a histologic marker of thyroid cancer. However, the pharmacological lectin-based approach has not been well studied. In the present study, we aimed to investigate the therapeutic potential of novel galectin-3 inhibitors by treating thyroid
Martijn J C van der Lienden et al.
International journal of molecular sciences, 22(5) (2021-04-04)
The lysosomal storage disease Niemann-Pick type C (NPC) is caused by impaired cholesterol efflux from lysosomes, which is accompanied by secondary lysosomal accumulation of sphingomyelin and glucosylceramide (GlcCer). Similar to Gaucher disease (GD), patients deficient in glucocerebrosidase (GCase) degrading GlcCer
Malene Laage Ebstrup et al.
Frontiers in cell and developmental biology, 11, 1211498-1211498 (2024-02-13)
Lysosomes are crucial organelles essential for various cellular processes, and any damage to them can severely compromise cell viability. This study uncovers a previously unrecognized function of the calcium- and phospholipid-binding protein Annexin A7 in lysosome repair, which operates independently
Juan García-Revilla et al.
Acta neuropathologica, 146(1), 51-75 (2023-05-19)
Parkinson's Disease (PD) is a neurodegenerative and progressive disorder characterised by intracytoplasmic inclusions called Lewy bodies (LB) and degeneration of dopaminergic neurons in the substantia nigra (SN). Aggregated α-synuclein (αSYN) is known to be the main component of the LB.
Kevin Burbidge et al.
Autophagy, 18(5), 1020-1048 (2021-10-07)
Numerous lines of evidence support the premise that the misfolding and subsequent accumulation of SNCA/α-synuclein (synuclein alpha) is responsible for the underlying neuronal pathology observed in Parkinson disease (PD) and other synucleinopathies. Moreover, the cell-to-cell transfer of these misfolded SNCA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service