Skip to Content
Merck
All Photos(2)

Key Documents

MABN2268

Sigma-Aldrich

Anti-LRRK1 Antibody, clone 12F2.1

clone 12F2.1, from mouse

Synonym(s):

Leucine-rich repeat serine/threonine-protein kinase 1, Leucine-rich repeat kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12F2.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... LRRK1(79705)

Related Categories

General description

Leucine-rich repeat serine/threonine-protein kinase 1 (UniProt: Q38SD2, Leucine-rich repeat kinase 1) is encoded by the LRRK1 (also known as KIAA1790) gene (Gene ID: 79705) in human. Leucine-rich repeat kinase 1 is a member of the ROCO family of protein kinases. Two isoforms of Leucine-rich repeat kinase 1 have been described that are generated by alternative splicing. It contains a ROC (Ras in complex proteins) domain together with a MAPKKK-like kinase domain and several protein protein interactions. It is reported to form a complex with activated EGFR via interaction with Grb2. Subsequently, it is internalized to early endosomes along with EGFR. Leucine-rich repeat kinase 1 also regulates EGFR transport from early to late endosomes and is involved in regulating the motility of EGF-containing early endosomes that is dependent on its kinase activity. Leucine-rich repeat kinase 1 is also reported to play a role in regulating bone mass and mice with defective gene are prone to osteoporosis, increased bone mineralization, and diminished bone resorption. (Ref.: Hanafusa, H et al. (2011). Nature Commun. 2, Article number: 158).

Specificity

Clone 12F2.1 detects Leucine-rich repeat serine/threonine-protein kinase 1 (LRRK1) in human. It targets an epitope within 86 amino acids from the internal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 86 amino acids from the internal region of human Leucine-rich repeat serine/threonine-protein kinase 1.

Application

Anti-LRRK1, clone 12F2.1, Cat. No. MABN2268, is a highly specific mouse monoclonal antibody that detects Leucine-rich repeat serine/threonine-protein kinase 1 (LRRK1) and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected LRRK1 in human cerebral cortex, human testis, and human pancreatic cancer tissues.

Quality

Evaluated by Western Blotting in human hypothalamus tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected LRRK1 in 10 µg of human hypothalamus tissue lysate.

Target description

~200 kDa observed; 225.39 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service