MABF424
Anti-CD44 Antibody (human/mouse), APC-Cy7, clone IM7
clone IM7, 0.2 mg/mL, from rat
Synonym(s):
CD44 antigen, Extracellular matrix receptor III, ECMR-III, GP90 lymphocyte homing/adhesion receptor, HUTCH-I, Hermes antigen, Hyaluronate receptor, Lymphocyte antigen 24, Ly-24, Phagocytic glycoprotein 1, PGP-1, Phagocytic glycoprotein I, PGP-I, CD44
About This Item
biological source
rat
Quality Level
conjugate
APC-Cy7
antibody form
purified antibody
antibody product type
primary antibodies
clone
IM7, monoclonal
species reactivity
human, mouse
concentration
0.2 mg/mL
technique(s)
flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG2bκ
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... CD44(960)
mouse ... Cd44(12505)
General description
Immunogen
Application
Inflammation & Immunology
Quality
Flow Cytometry Analysis: 0.25 μg of this antibody detected CD44 in one million C57Bl/6 splenocytes.
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service