Skip to Content
Merck
All Photos(1)

Key Documents

MABE341

Sigma-Aldrich

Anti-Puromycin Antibody, clone 17H1

clone 17H1, from rat

Synonym(s):

Clone 17H1 Anti-Puromycin, Puromycin Detection Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

17H1, monoclonal

species reactivity

human

species reactivity (predicted by homology)

all

technique(s)

western blot: suitable

isotype

IgG2aκ

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NPEPPS(9520)

General description

Puromycin is an aminonucleoside antibiotic, derived from the Streptomyces alboniger bacterium, that functions as a protein synthesis inhibitor that blocks translation through premature chain termination in the ribosome. Monoclonal antibodies to puromycin may be used with standard immunochemical methods to directly monitor translation, a method known as surface sensing of translation (SUnSET). Part of the molecule resembles the 3′ end of the aminoacylated tRNA, making it useful for protein translation analysis. Puromycin induces DNA fragmentation in thymocytes and in human HL-60 leukemia cells.

Specificity

Demonstrated to react with Human test sample, preincubated with Puromycin. Predicted to react with all species when test sample is incubated with Puromycin.

Immunogen

Puromycin from Streptomyces alboniger

Application

Anti-Puromycin antibody, clone 17H1 detects puromycin incorporated into protein. Monoclonal antibodies to puromycin may also be used with standard immunochemical methods.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins

Quality

Evaluated by Western Blotting in HEK293 cell lysates treated with Puromycin and Cyclohexamide, or with Puromycin only.

Western Blotting Analysis: A 1:100,000 dilution of this antibody detected Puromycin-incorporated neosynthesized proteins in HEK293 cell lysates treated with Puromycin only. This antibody also detected small mounts of Puromycin-incorporated neosynthesized proteins in HEK293 cells treated with Puromycin and Cyclohexamide.

Target description

Puromycin is incorporated in neosynthesized proteins. In the presense of Puromycin only, this antibody detects Puromycin-incorporated neosynthesized proteins at multiple molecular weights. However, a decrease in signal is observed in the co-presense of Cycloheximide, an inhibitor of protein biosynthesis in eukaryotic organisms.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysates treated with Puromycin and Cyclohexamide, or with Puromycin only.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Faraz Ahmad et al.
Bioscience reports, 38(3) (2018-04-28)
Neonatal neuropsychiatric stress induces alterations in neurodevelopment that can lead to irreversible damage to neuronal physiology, and social, behavioral, and cognitive skills. In addition, this culminates to an elevated vulnerability to stress and anxiety later in life. Developmental deficits in
Faraz Ahmad et al.
Neuropsychiatric disease and treatment, 14, 3289-3298 (2018-12-21)
Lead (Pb) is a persistent environmental neurotoxin and its exposure even in minute quantities has been known to induce neuronal defects. The immature brain is singularly sensitive to Pb neurotoxicity, and its exposure during development has permanent detrimental effects on
Yizhe Sun et al.
Nature communications, 12(1), 5147-5147 (2021-08-28)
PTEN is frequently mutated in human cancers and PTEN mutants promote tumor progression and metastasis. PTEN mutations have been implicated in immune regulation, however, the underlying mechanism is largely unknown. Here, we report that PTENα, the isoform of PTEN, remains
Zhongyan Wu et al.
EMBO reports, 24(5), e55641-e55641 (2023-03-18)
Stress granules are dynamic cytoplasmic ribonucleoprotein granules that assemble in response to cellular stress. Aberrant formation of stress granules has been linked to neurodegenerative diseases. However, the molecular mechanisms underlying the initiation of stress granules remain elusive. Here we report
Hong Sun et al.
Aging, 12(20), 20432-20444 (2020-10-29)
In human macrophages and monocytes, lipopolysaccharide (LPS) induces nuclear factor kappa B (NFκB) activation and pro-inflammatory cytokines production. We tested the possible involvement of Ku70 and Ku80 in the process. In THP-1 macrophages and primary human peripheral blood mononuclear cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service