Skip to Content
Merck
All Photos(1)

Key Documents

MABC39

Sigma-Aldrich

Anti-LAMP-1 (CD107a) Antibody, clone 1D4B

clone 1D4B, from rat

Synonym(s):

lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1, CD107 antigen-like family member A, CD107a antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D4B, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Lamp1(16783)

General description

Lysosome-associated membrane protein 1 (LAMP-1) is a highly glycosolated, single-pass type 1 membrane protein belonging to the lysosome- associated membrane protein family; a group of proteins that make up the majority of the lysosomal membrane and are considered crucial mediators for cellular adhesion by interacting with carbohydrate receptors. LAMP-1 functions to traffic carbohydrate ligands to selectins. It is also thought to play a role in tumor cell metastasis.

Immunogen

NIH/3T3 plasma membrane fraction of mouse LAMP-1.

Application

Detect LAMP-1 (CD107a) using this Anti-LAMP-1 (CD107a) Antibody, clone 1D4B validated for use in WB, IP, IC, IH, FC.
Immunoprecipitation Analysis: A previous lot was used by an independent laboratory in IP (Chen, 1985).

Immunocytochemistry Analysis: A previous lot was used by an independent laboratory in IC (Chen, 1985).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Tumor Markers

Quality

Evaluated by Western Blot in NIH/3T3 cell lysate.

Western Blot Analysis: 1:50,000 dilution of this antibody detected LAMP-1 in 10 µg of NIH/3T3 cell lysate.

Target description

~ 100 kDa observed. The calculated molecular weight of this protein is 44 kDa The protein is heavily glycosolated and therefore its molecular weight is typically noted from 95-115 kDa

Physical form

Format: Purified
Protein G
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
NIH/3T3 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pablo Izquierdo et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(42) (2021-10-14)
Microglia are the resident immune cells of the central nervous system. They constantly survey the brain parenchyma for redundant synapses, debris, or dying cells, which they remove through phagocytosis. Microglial ramification, motility, and cytokine release are regulated by tonically active
Christian Vogl et al.
The EMBO journal, 35(23), 2536-2552 (2016-07-28)
The transmembrane recognition complex (TRC40) pathway mediates the insertion of tail-anchored (TA) proteins into membranes. Here, we demonstrate that otoferlin, a TA protein essential for hair cell exocytosis, is inserted into the endoplasmic reticulum (ER) via the TRC40 pathway. We
Jeong-Sun Ju et al.
The journal of physiological sciences : JPS, 66(5), 417-430 (2016-03-05)
Mitochondrial homeostasis is tightly regulated by two major processes: mitochondrial biogenesis and mitochondrial degradation by autophagy (mitophagy). Research in mitochondrial biogenesis in skeletal muscle in response to endurance exercise training has been well established, while the mechanisms regulating mitophagy and
Urmi Bandyopadhyay et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(30), 11055-11060 (2014-07-16)
Lipofuscin, or aging pigment, is accreted as red autofluorescence in the lysosomes of motor neuron cell bodies in the ventral horn of WT mice by 3 mo of age. Strikingly, in two presymptomatic ALS mouse strains transgenic for mutant human
Paulina Kaminska et al.
EMBO reports, 25(5), 2278-2305 (2024-03-19)
SorLA, encoded by the gene SORL1, is an intracellular sorting receptor of the VPS10P domain receptor gene family. Although SorLA is best recognized for its ability to shuttle target proteins between intracellular compartments in neurons, recent data suggest that also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service