Skip to Content
Merck
All Photos(1)

Key Documents

MAB930

Sigma-Aldrich

Anti-Staphylococcus aureus Antibody, clone STAPH 11-248.2

ascites fluid, clone STAPH 11-248.2, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

STAPH 11-248.2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

isotype

IgM

shipped in

wet ice

Specificity

Staphylococcus aureus.

No cross-reactivity with Streptococcus sp., Mycoplasma sp., or Corynebacteria sp.

Immunogen

UV-inactivated Staphylococcus aureas cells (ATCC# 29740).

Application

Detect Staphylococcus aureus using this Anti-Staphylococcus aureus Antibody, clone STAPH 11-248.2 validated for use in ELISA.
ELISA at 1:16,000 using 19.0 mg/mL

Optimal working dilution must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Bacterial

Physical form

Ascites fluid. Liquid containing no preservatives.

Storage and Stability

Maintain at 2-8°C in convenient aliquots for up to 12 months from date of receipt.

During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200μl or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

J Ou et al.
Rhinology, 55(4), 305-311 (2017-07-09)
Staphylococcus aureus (S. aureus) can reside within the sinonasal mucosa in chronic rhinosinusitis patients and causes recurrent infections. Within the host cell, S. aureus can evade host immune detection enabling its own survival. We hypothesise that S. aureus can persist
Judy Ou et al.
International forum of allergy & rhinology, 6(8), 792-799 (2016-04-16)
Staphylococcus aureus (S. aureus) has been shown to exist within nasal epithelial cells in chronic rhinosinusitis (CRS) patients. This study investigates the localization of intracellular S. aureus (ICSA) in CRS patients, the associated histopathology changes, and their effect on long-term
R S Root-Bernstein
Medical hypotheses, 44(1), 20-27 (1995-01-01)
Several investigators have proposed that autoimmunity may be induced by idiotype-antiidiotype antibody networks. It is generally assumed that the antiidiotype is produced in response to the idiotype, and therefore that autoimmune diseases have single antigenic initiators. The theory of multiple-antigen-mediated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service